Merck
  • Treatment with FLT3 inhibitor in patients with FLT3-mutated acute myeloid leukemia is associated with development of secondary FLT3-tyrosine kinase domain mutations.
Treatment with FLT3 inhibitor in patients with FLT3-mutated acute myeloid leukemia is associated with development of secondary FLT3-tyrosine kinase domain mutations.
Cancer (2014-04-17)
Yesid Alvarado, Hagop M Kantarjian, Rajyalakshmi Luthra, Farhad Ravandi, Gautam Borthakur, Guillermo Garcia-Manero, Marina Konopleva, Zeev Estrov, Michael Andreeff, Jorge E Cortes
ABSTRACT
FLT3-internal tandem duplication (ITD) mutations are found in approximately 30% of patients with acute myeloid leukemia (AML). FLT3 inhibitors have shown clinical activity in AML with FLT3-ITD, but responses are usually short-lived. This study examined 69 FLT3-mutated patients with AML, who were treated with different FLT3 inhibitors to analyze emergence of new mutations. At baseline, 87% of patients had an ITD mutation, 7% had a D835/I836 mutation, and 6% had combined ITD and D835/I836 mutations. Responses occurred in 32% of patients, all with FLT3-ITD; none of the patients with D835/I836 or ITD+D835/I836 responded. Mutational assessment at the time of FLT3 inhibitor discontinuation showed that 68% of patients were unchanged, 10% had become undetectable, and 22% of patients progressed from a single ITD to have combined ITD+D835/I836 mutations. In those patients with unchanged FLT3 mutation at progression, the median survival was 5 months, whereas in those with undetectable and with combined ITD+D835/I836 mutations, the median survival was 7 months, respectively. These data confirm in vitro observations that a secondary tyrosine kinase domain mutation may arise after the use of FLT3 inhibitors in patients with single FLT3-ITD mutated AML, a phenomenon that is associated with resistance and a poor prognosis.
Sigma-Aldrich
FLT3 (571-993), active, GST tagged human, PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Supelco
Nicotinamide (Niacinamide), analytical standard
Sigma-Aldrich
Nicotinamide, ≥99.5% (HPLC)
Sigma-Aldrich
Nicotinamide, ≥98.5% (HPLC)
Sigma-Aldrich
Niacinamide, meets USP testing specifications
Sigma-Aldrich
Nicotinamide, ≥98% (HPLC), powder
Sigma-Aldrich
Nicotinamide, BioReagent, suitable for cell culture, suitable for insect cell culture
Supelco
Niacinamide, Pharmaceutical Secondary Standard; Certified Reference Material
Nicotinamide, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicotinamide (Vitamin B3) solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
USP
Niacinamide, United States Pharmacopeia (USP) Reference Standard