Merck
  • Influence of stearic acid on cholesterol metabolism relative to other long-chain fatty acids.
Influence of stearic acid on cholesterol metabolism relative to other long-chain fatty acids.
The American journal of clinical nutrition (1994-12-01)
S M Grundy
ABSTRACT
Stearic acid is a long-chain saturated fatty acid. However, in contrast with other saturated fatty acids, stearic acid apparently does not raise serum cholesterol concentrations. Studies carried out three decades ago provided strong suggestive evidence that this was the case. More recent investigations that specifically compared stearic acid with other fatty acids in human studies have confirmed that stearic acid is not hypercholesterolemic. Stearic acid was shown not to raise low-density-lipoprotein cholesterol relative to oleic acid, which is known to be neutral in its effects on cholesterol concentrations. In contrast, palmitic acid, another long-chain saturated fatty acid, definitely raises cholesterol concentrations. For this reason, fats rich in stearic acid might be used in place of those high in palmitic acid in cholesterol-lowering diets.
Product Description
Supelco
Stearic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Stearic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Stearic acid, analytical standard
Sigma-Aldrich
Stearic acid, reagent grade, 95%
Sigma-Aldrich
Stearic acid, ≥95%, FCC, FG
Stearic acid, European Pharmacopoeia (EP) Reference Standard
USP
Stearic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Stearic acid, Grade I, ≥98.5% (capillary GC)