Merck
  • [Use of gas chromatography for determining benzoic and sorbic acid levels in orange beverage and tomato concentrate].
[Use of gas chromatography for determining benzoic and sorbic acid levels in orange beverage and tomato concentrate].
Roczniki Panstwowego Zakladu Higieny (1990-01-01)
H Giryn, Z Gruszczyńska
ABSTRACT
Conditions of simultaneous determination of benzoic and sorbic acid in orange drink and tomato concentrate by gas chromatography method were developed and tested. Preservatives were isolated by extraction of the samples with diethyl ether in acidic media, and then converted into methyl esters with thionyl chloride. Methyl esters were separated on column packed with 15% EGA coated on 80-100 mesh Chromosorb W AW DMCS. The smallest amount of preservatives detectable on chromatography column is 20 ng for benzoic acid and 16 ng for sorbic acid. The recovery rate of the method ranged from 96-97% for benzoic acid and 89-92% for sorbic acid, while the variability coefficient was from 1.7-3.5% for benzoic acid and 4.5-6% for sorbic acid.
Sigma-Aldrich
Sorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
Sorbic acid, 99.0-101.0% anhydrous basis
Sorbic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Sorbic acid, Pharmaceutical Secondary Standard; Certified Reference Material