Merck
The pathogenesis of cystinosis: mechanisms beyond cystine accumulation.
American journal of physiology. Renal physiology (2010-09-10)
Martijn J Wilmer, Francesco Emma, Elena N Levtchenko
ABSTRACT
Renal proximal tubules are highly sensitive to ischemic and toxic insults and are affected in diverse genetic disorders, of which nephropathic cystinosis is the most common. The disease is caused by mutations in the CTNS gene, encoding the lysosomal cystine transporter cystinosin, and is characterized by accumulation of cystine in the lysosomes throughout the body. In the majority of the patients, this leads to generalized proximal tubular dysfunction (also called DeToni-Debré-Fanconi syndrome) in the first year and progressive renal failure during the first decade. Extrarenal organs are affected by cystinosis as well, with clinical symptoms manifesting mostly after 10 yr of age. The cystine-depleting agent cysteamine significantly improves life expectancy of patients with cystinosis, but offers no cure, pointing to the complexity of the disease mechanism. In this review, current knowledge on the pathogenesis of cystinosis is described and placed in perspective of future research.
Product Description
Supelco
L-Cystine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Cystine
Supelco
L-Cystine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Cystine, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.5%
Cystine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Cystine, ≥99.7% (TLC)
Sigma-Aldrich
L-Cystine, ≥98% (TLC), crystalline
Sigma-Aldrich
L-Cystine, from non-animal source, meets EP testing specifications, suitable for cell culture, 98.5-101.0%