Skip to Content
Merck
All Photos(1)

Key Documents

30200

Sigma-Aldrich

L-Cystine

≥99.7% (TLC)

Synonym(s):

(R,R)-3,3′-Dithiobis(2-aminopropionicacid)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-SCH2CH(NH2)CO2H]2
CAS Number:
Molecular Weight:
240.30
Beilstein:
1728094
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Cystine, ≥99.7% (TLC)

Quality Level

Assay

≥99.7% (TLC)

form

powder or crystals

optical activity

[α]20/D −219±5°, c = 1% in 1 M HCl

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying

color

white

mp

>240 °C (dec.) (lit.)

solubility

1 M HCl: 0.5 g/10 mL, clear, colorless

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤100 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤6 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤150 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

peptide synthesis

SMILES string

N[C@@H](CSSC[C@H](N)C(O)=O)C(O)=O

InChI

1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1

InChI key

LEVWYRKDKASIDU-IMJSIDKUSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Cystine has been used to prepare stock solution which in turn is utilized for determining the L-cysteine and L-cystine in pharmaceutical preparations as well as in urine samples by using spectrofluorimetric flow injection method.

Biochem/physiol Actions

Cystine is required for the synthesis of glutathione that protects the cells from oxidative stress. A specific x(c)-cysteine/glutamate antiporter mediates cellular uptake of cysteine in exchange for glutamate. Inhibition of cysteine uptake induces toxicity in neuronal cells.

Other Notes

Preparation of N,N′-dibenzoyl-L-cystine, an efficient auxiliary for enantioselective reductions of ketones

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 4

1 of 4

T H Murphy et al.
Neuron, 2(6), 1547-1558 (1989-06-01)
Glutamate binds to both excitatory neurotransmitter binding sites and a Cl(-)-dependent, quisqualate- and cystine-inhibited transport site on brain neurons. The neuroblastoma-primary retina hybrid cells (N18-RE-105) are susceptible to glutamate-induced cytotoxicity. The Cl(-)-dependent transport site to which glutamate and quisqualate (but
Journal of the Chemical Society. Chemical Communications, 801-801 (1987)
T Pérez-Ruiz et al.
The Analyst, 117(6), 1025-1028 (1992-06-01)
A flow injection configuration is proposed for the determination of L-cysteine and L-cystine individually and for mixtures of both analytes. The procedure is based on the rapid oxidation of L-cysteine by thallium(III) with concomitant formation of fluorescent thallium(I). The inclusion
K. Soai et al.
Journal of the Chemical Society. Chemical Communications, 413-413 (1984)
Philip J Hogg
Nature reviews. Cancer, 13(6), 425-431 (2013-05-11)
Protein action in nature is generally controlled by the amount of protein produced and by chemical modification of the protein, and both are often perturbed in cancer. The amino acid side chains and the peptide and disulphide bonds that bind

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service