Merck
  • Age-related reference values for urinary organic acids in a healthy Turkish pediatric population.
Age-related reference values for urinary organic acids in a healthy Turkish pediatric population.
Clinical chemistry (1994-06-01)
F Guneral, C Bachmann
ABSTRACT
Organic acid concentrations were quantified by gas chromatography and the individual acids identified by mass spectrometry in urine specimens from a healthy Turkish pediatric population of ages 2 days to 16 years, subdivided into five age groups. We quantified 69 organic acids (32 major and 37 minor components) and report here the median values and percentiles for each compound, adjusted for creatinine content, for the five groups. Concentrations of most of the organic acids tend to decrease with age but display substantial differences between age groups. This emphasizes the importance of comparing patients' data with age-matched reference data. Correlations between the excretion of organic acids and protein or caloric intake were significant for several compounds.
Sigma-Aldrich
Terephthalic acid, 98%
Sigma-Aldrich
p-Cresol, ≥99%, FG
Sigma-Aldrich
L-(+)-Lactic acid solution, ~40% in H2O
Sigma-Aldrich
L-(+)-Lactic acid solution, meets analytical specification of Ph. Eur., BP, 88-92% total acid basis
Supelco
4-Methylphenol solution, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Glycolic acid, ReagentPlus®, 99%
Sigma-Aldrich
Phthalic acid, reagent grade, 98%
Sigma-Aldrich
2-Furoic acid, 98%
Sigma-Aldrich
3-Methylglutaric acid, 99%
Sigma-Aldrich
Glycolic acid solution, high purity, 70 wt. % in H2O
Sigma-Aldrich
Glycolic acid solution, technical grade, 70 wt. % in H2O
Sigma-Aldrich
Glutaric acid, 99%
Sigma-Aldrich
α-Hydroxyisobutyric acid, 98%
Sigma-Aldrich
3-Hydroxybutyric acid, 95%
Sigma-Aldrich
L-Pyroglutamic acid, ≥99.0% (T)
Sigma-Aldrich
Phthalic acid, puriss. p.a., ≥99.5% (T)
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
Sigma-Aldrich
4-Hydroxyphenylacetic acid, 98%
Sigma-Aldrich
L-(+)-Lactic acid solution, in H2O, ≥85%
Sigma-Aldrich
Phthalic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Methylsuccinic acid, 99%
Sigma-Aldrich
Pimelic acid, 98%
Sigma-Aldrich
α-Hydroxyisobutyric acid, 99%
Sigma-Aldrich
cis-Aconitic acid, ≥98%
Sigma-Aldrich
α-Ketoglutaric acid, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
α-Ketoglutaric acid, ≥98.5% (NaOH, titration)
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
L-Pyroglutamic acid, BioXtra
Sigma-Aldrich
Glycolic acid, BioXtra, ≥98.0% (titration)