Merck
  • Degrees of freedom effect on fragmentation in tandem mass spectrometry of singly charged supramolecular aggregates of sodium sulfonates.
Degrees of freedom effect on fragmentation in tandem mass spectrometry of singly charged supramolecular aggregates of sodium sulfonates.
Journal of mass spectrometry : JMS (2013-03-16)
Serena Indelicato, David Bongiorno, Sergio Indelicato, Laszlo Drahos, Vincenzo Turco Liveri, Lilla Turiák, Karoly Vékey, Leopoldo Ceraulo
ABSTRACT
The characteristic collision energy (CCE) to obtain 50% fragmentation of positively and negatively single charged noncovalent clusters has been measured. CCE was found to increase linearly with the degrees of freedom (DoF) of the precursor ion, analogously to that observed for synthetic polymers. This suggests that fragmentation behavior (e.g. energy randomization) in covalent molecules and clusters are similar. Analysis of the slope of CCE with molecular size (DoF) indicates that activation energy of fragmentation of these clusters (loss of a monomer unit) is similar to that of the lowest energy fragmentation of protonated leucine-enkephalin. Positively and negatively charged aggregates behave similarly, but the slope of the CCE versus DoF plot is steeper for positive ions, suggesting that these are more stable than their negative counterparts.
Product Description
Supelco
Sodium 1-butanesulfonate, suitable for ion pair chromatography, LiChropur, ≥99.0% (T)
Sigma-Aldrich
Sodium methanesulfonate, 98%
Sigma-Aldrich
Potassium methanesulfonate, ≥98.0% (dry substance, T)
Sigma-Aldrich
Sodium 1-butanesulfonate, 98%
Sigma-Aldrich
Sodium 1-butanesulfonate, anionic detergent
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, ≥98%
Sigma-Aldrich
1-Octanesulfonic acid sodium salt, BioXtra
Supelco
Methanesulfonic acid concentrate, 0.1 M CH3SO3H in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Silver methanesulfonate
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)