Skip to Content
Merck
All Photos(4)

Key Documents

O0133

Sigma-Aldrich

1-Octanesulfonic acid sodium salt

BioXtra

Synonym(s):

Octane-1-sulfonic acid sodium salt, Sodium 1-octanesulfonate, Sodium octane-1-sulfonate, Sodium octanesulfonate, n-octyl sulfate, sodium 1-octanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C8H17O3SNa
CAS Number:
Molecular Weight:
216.27
EC Number:
MDL number:
UNSPSC Code:
12161900
PubChem Substance ID:
NACRES:
NA.21

description

anionic

Quality Level

product line

BioXtra

mol wt

216.27 g/mol

technique(s)

HPLC: suitable

impurities

≤0.005% Phosphorus (P)
≤0.1% Insoluble matter

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤0.05%

cation traces

Al: ≤0.0005%
Ca: ≤0.001%
Cu: ≤0.0005%
Fe: ≤0.0005%
K: ≤0.005%
Mg: ≤0.0005%
NH4+: ≤0.05%
Pb: ≤0.001%
Zn: ≤0.0005%

SMILES string

[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na/c1-2-3-4-5-6-7-8-12(9,10)11;/h2-8H2,1H3,(H,9,10,11);/q;+1/p-1

InChI key

HRQDCDQDOPSGBR-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ion-pairing reagent for HPLC analysis of peptides and proteins.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P G Jaison et al.
Journal of chromatography. A, 1216(9), 1383-1389 (2009-01-27)
Studies were carried out for the separation of uranium (U) and thorium (Th) on reversed-phase (RP) C18 columns using mandelic acid as an eluent. Retention of thorium-mandelate on the unmodified stationary phase was found to be greater than that of
I T James et al.
Journal of chromatography, 525(1), 43-57 (1990-01-26)
Following a detailed study, a rapid and sensitive assay for the naturally fluorescent collagen cross-links pyridinoline and deoxypyridinoline has been developed using ion-pair reversed-phase high-performance liquid chromatography in the presence of 1-octanesulphonic acid (OSA). Pyridinoline and deoxypyridinoline were separated on
Tejwant Singh et al.
Physical chemistry chemical physics : PCCP, 12(37), 11728-11735 (2010-08-10)
We have observed dual transitions in various physical properties while investigating the surfactant-like behavior of the ionic liquid (IL) 1-butyl-3-methylimidazolium octylsulfate [C(4)mim][C(8)SO(4)] in aqueous media. Interestingly, in conductivity measurements, it was found that the second transition, which is comparatively weak
J L Razak et al.
Pharmaceutical research, 18(1), 104-111 (2001-05-05)
The aim of this study was to develop a method based upon electrokinetic chromatography (EKC) using oppositely charged surfactant vesicles as a buffer modifier to estimate hydrophobicity (log P) for a range of neutral and charged compounds. Vesicles were formed
Yingjie Ma et al.
Chemical communications (Cambridge, England), 47(45), 12340-12342 (2011-10-21)
By the introduction of trimethylammonium groups at both upper and lower rims, a cationic water-soluble pillar[5]arene was prepared, which forms a stable 1 : 1 host-guest complex with sodium 1-octanesulfonate in water.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service