Skip to Content
Merck
All Photos(3)

Key Documents

304506

Sigma-Aldrich

Sodium methanesulfonate

98%

Synonym(s):

Methanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3SO3Na
CAS Number:
Molecular Weight:
118.09
Beilstein:
3915209
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

solid

SMILES string

[Na+].CS([O-])(=O)=O

InChI

1S/CH4O3S.Na/c1-5(2,3)4;/h1H3,(H,2,3,4);/q;+1/p-1

InChI key

KKVTYAVXTDIPAP-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Sodium methanesulfonate is generally used as an anion source [CH3SO3] for synthesizing ionic liquids (PILs).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitrile-functionalized azepanium ionic liquids: Synthesis characterization and thermophysical properties.
Lethesh K C, et al.
Journal of Molecular Liquids, 221, 1140-1144 (2016)
Antonios Pantazis et al.
The Journal of physiology, 598(22), 5245-5269 (2020-08-25)
KV 1.2 channels, encoded by the KCNA2 gene, regulate neuronal excitability by conducting K+ upon depolarization. A new KCNA2 missense variant was discovered in a patient with epilepsy, causing amino acid substitution F302L at helix S4, in the KV 1.2
Sunghoon Kim et al.
Nature communications, 10(1), 3703-3703 (2019-08-20)
A family of plant nuclear ion channels, including DMI1 (Does not Make Infections 1) and its homologs CASTOR and POLLUX, are required for the establishment of legume-microbe symbioses by generating nuclear and perinuclear Ca2+ spiking. Here we show that CASTOR
Novel imidazolium-based poly (ionic liquid) s with different counterions for self-healing.
Cui J, et al.
Journal of Material Chemistry A, 5(48), 25220-25229 (2017)
Laura Mortara et al.
Langmuir : the ACS journal of surfaces and colloids, 34(37), 11049-11057 (2018-08-22)
Zwitterionic micelles adsorb anions and several techniques were used to determine the specificity of this interaction. Although at a lower intensity, this adsorption can be compared to those observed in cationic micelles, which showed that interfacial dehydration is a fundamental

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service