Merck
  • Pyrophosphate sensing by a fluorescent Zn2+ bound triazole linked imino-thiophenyl conjugate of calix[4]arene in HEPES buffer medium: spectroscopy, microscopy, and cellular studies.
Pyrophosphate sensing by a fluorescent Zn2+ bound triazole linked imino-thiophenyl conjugate of calix[4]arene in HEPES buffer medium: spectroscopy, microscopy, and cellular studies.
Analytical chemistry (2012-05-04)
Rakesh K Pathak, Khatija Tabbasum, Ankit Rai, Dulal Panda, Chebrolu P Rao
ABSTRACT
An in situ prepared Zn(2+) complex of triazole linked imino-thiophenyl conjugate of calix[4]arene, [ZnL], was demonstrated to be highly fluorescent in HEPES buffer solution. [ZnL] has been used as a chemo-sensing ensemble for the recognition of phosphates in general and pyrophosphates in particular among the eighteen different anions studied. The chemo-sensing behavior of the [ZnL] has been demonstrated through fluorescence, absorption, visual fluorescent color changes, ESI MS, and (1)H NMR titrations. Variations in the microstructural features of L, its zinc complex and the complex upon addition of PPi have been demonstrated through atomic force microscopy and transmission electron microscopy. Such studies have been extended to see the permeability of the conjugate into the HeLa cells by fluorescence microscopy. In accession, a reversible "write-read-erase-read" logic gate property of L has been demonstrated through a feedback loop in the presence of Zn(2+) and PPi.
Sigma-Aldrich
HEPES, BioUltra, for molecular biology, ≥99.5% (T)
Sigma-Aldrich
HEPES sodium salt solution, BioReagent, 1M, suitable for cell culture
Sigma-Aldrich
HEPES sodium salt, ≥99.0% (titration)
Sigma-Aldrich
HEPES sodium salt, BioPerformance Certified, suitable for cell culture, ≥99.0%
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
SAFC
HEPES
SAFC
HEPES sodium salt
Sigma-Aldrich
HEPES sodium salt, ≥99.5% (titration), free-flowing, Redi-Dri
Sigma-Aldrich
HEPES solution, BioPerformance Certified, 1 M, suitable for cell culture, 0.2 μm filtered
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
HEPES solution, 1 M, pH 7.0-7.6, sterile-filtered, BioReagent, suitable for cell culture