Skip to Content
Merck
All Photos(5)

Key Documents

H3784

Sigma-Aldrich

HEPES sodium salt

BioPerformance Certified, suitable for cell culture, ≥99.0%

Synonym(s):

4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N′-(2-ethanesulfonic acid) sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H17N2NaO4S
CAS Number:
Molecular Weight:
260.29
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:
NACRES:
NA.25

grade

BioPerformance Certified

Quality Level

Assay

≥99.0%

form

powder

storage condition

dry at room temperature

technique(s)

activity assay: suitable
cell culture | mammalian: suitable

impurities

endotoxin and total aerobic microbial count, tested

color

white

pH

10-12 (1 M in water)

useful pH range

6.8-8.2

pKa (25 °C)

7.5

solubility

water: 0.1 M, clear, colorless

density

1.19 g/cm3 at 20—25 °C
1.21 g/cm3 at 20—25 °C

absorption

≤0.02 at 280 in H2O at 0.1 M
≤0.04 at 260 in H2O at 0.1 M

suitability

suitable for component for culture media
suitable for molecular biology

application(s)

cell analysis
diagnostic assay manufacturing
general analytical
life science and biopharma

foreign activity

DNase, RNase, NICKase, protease, none detected

SMILES string

[Na+].OCCN1CCN(CC1)CCS([O-])(=O)=O

InChI

1S/C8H18N2O4S.Na/c11-7-5-9-1-3-10(4-2-9)6-8-15(12,13)14;/h11H,1-8H2,(H,12,13,14);/q;+1/p-1

InChI key

RDZTWEVXRGYCFV-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

HEPES, or 4-(2-hydroxyethyl)piperazine-1-ethane-sulfonic acid, is a zwitterionic N-substituted aminosulfonic acid buffer with a remarkable pH buffering capacity between 6.8 and 8.2 (pKa of 7.48 at 25°C). Widely recognized for its versatility, HEPES is extensively employed in cell biology, biochemical, and biological research. Notably, its neutrality towards metal ions makes it a preferred choice for solutions with metal ions, acting as a "non-coordinating buffer."

In cell culture, HEPES excels in maintaining stable physiological pH even amidst fluctuating carbon dioxide concentrations, outperforming bicarbonate buffers (NaHCO3). Moreover, HEPES showcases valuable properties in various biological and biochemical processes, such as isoelectric focusing for protein separation and minimal interference in DNA-restriction enzyme reactions, making it indispensable across diverse research applications, including immunoprecipitation, cell lysis, and live cell imaging. Its versatility, exceptional buffering capacity, and minimal interaction with molecules establish HEPES as a key tool in various research domains.

Application

HEPES sodium salt has been used:
  • to supplement the HSOF medium
  • as a buffer component of HCZB medium in egg and embryo culture
  • as a buffer component in cell culture

Features and Benefits

  • High-purity product for biochemical and biological research
  • Tested to confirm low levels of heavy metal contamination, ensuring suitability for various applications
  • Highly soluble in water with a useful pH range of 6.8 - 8.2 and pKa of 7.5 at 25 °C
  • Suitable for Cell culture
  • Tested for Endotoxins and Total Aerobic Microbial Count
  • Free from DNase, NICKase, RNase, Endonuclease, Exonuclease and Protease

Physical form

Lyophilized, spray dried, or crystallized material.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mingyue Li et al.
Structure (London, England : 1993), 27(5), 806-815 (2019-03-19)
The peroxidation of cardiolipins by reactive oxygen species, which is regulated and enhanced by cytochrome c (cyt c), is a critical signaling event in mitochondrial apoptosis. We probe the molecular underpinnings of this mitochondrial death signal through structural and functional
Ekaterina Isaac et al.
Theriogenology, 161, 273-284 (2020-12-29)
The growth of viable cattle embryos in culture to stages beyond the hatching blastocyst is of interest to developmental biologists wishing to understand developmental events beyond the first lineage decision, as well as for commercial applications, because a lengthening of
Ines Rosignol et al.
International journal of molecular sciences, 21(5) (2020-03-14)
Mitochondrial damage plays a prominent role in glaucoma. The only way cells can degrade whole mitochondria is via autophagy, in a process called mitophagy. Thus, studying mitophagy in the context of glaucoma is essential to understand the disease. Up to
Artem Babaian et al.
Cell reports, 31(5), 107611-107611 (2020-05-07)
The ribosome is an RNA-protein complex that is essential for translation in all domains of life. The structural and catalytic core of the ribosome is its ribosomal RNA (rRNA). While mutations in ribosomal protein (RP) genes are known drivers of
Jihye Park et al.
The Analyst, 137(19), 4411-4414 (2012-08-07)
We have developed a quickly responsive, and specific fluorescent assay for the detection of Au(III) on the basis of the formation of gold nanoparticles in the presence of HEPES, which cleave the C-I bond of I-BODIPY 1 to yield the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service