Merck
  • Direct synthesis of 6-arylpurines by reaction of 6-chloropurines with activated aromatics.
Direct synthesis of 6-arylpurines by reaction of 6-chloropurines with activated aromatics.
The Journal of organic chemistry (2010-08-12)
Hai-Ming Guo, Pu Li, Hong-Ying Niu, Dong-Chao Wang, Gui-Rong Qu
ABSTRACT
Highly functionalized C6-aryl-substituted purine analogues were synthesized through direct arylation of 6-chloropurine with aromatics promoted by anhydrous AlCl(3) in a single step. The reactions, which were conducted using a 3-fold excess of AlCl(3) in refluxing 1,2-dichloroethane, gave moderate to excellent product yields in 0.5 h. This work is complementary to the classical coupling reactions for the synthesis of C6-aryl-substituted purine analogues.
Sigma-Aldrich
6-Chloropurine, ≥99%
Sigma-Aldrich
6-Chloropurine, 99%