Merck
  • Cloning, sequence, and expression of the lysostaphin gene from Staphylococcus simulans.
Cloning, sequence, and expression of the lysostaphin gene from Staphylococcus simulans.
Proceedings of the National Academy of Sciences of the United States of America (1987-03-01)
P A Recsei, A D Gruss, R P Novick
ABSTRACT
A 1.5-kilobase-pair fragment of DNA that contains the lysostaphin gene from Staphylococcus simulans and its flanking sequences has been cloned and completely sequenced. The gene encodes a preproenzyme of Mr 42,000. The NH2-terminal sequence of the preproenzyme is composed of a signal peptide followed by seven tandem repeats of a 13-amino acid sequence. Conversion of prolysostaphin to the mature enzyme occurs extracellularly in cultures of S. simulans and involves removal of the NH2-terminal portion of the proenzyme that contains the tandem repeats. The high degree of homology of the repeats suggests that they have arisen by duplication of a 39-base-pair sequence of DNA. In S. simulans, the lysostaphin gene is present on a large beta-lactamase plasmid.
Sigma-Aldrich
Lysostaphin from Staphylococcus simulans, recombinant, expressed in E. coli, lyophilized powder
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, lyophilized powder, Protein 50-70 % by biuret, ≥500 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, aseptically filled
Sigma-Aldrich
Lysostaphin from Staphylococcus staphylolyticus, free of DNA contaminants, suitable for Microbiome research, lyophilized powder, ≥500 units/mg protein