Skip to Content
Merck
All Photos(1)

Key Documents

L9043

Sigma-Aldrich

Lysostaphin from Staphylococcus simulans

recombinant, expressed in E. coli, lyophilized powder

Synonym(s):

Glycyl-glycine Endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

biological source

bacterial

Quality Level

recombinant

expressed in E. coli

Assay

≥90% (HPLC)

form

lyophilized powder

specific activity

≥3000 unit/mg solid

technique(s)

molecular cloning: suitable

suitability

suitable for sequence analysis test

application(s)

life science and biopharma

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Lysostaphin, a zinc metalloenzyme, is mainly secreted as a proenzyme. It acts as an anti-staphylococcal agent and is highly stable at pH 4 and 5°C.

Application

Lysostaphin from Staphylococcus simulans has been used in a study to assess molecular cloning, sequencing, and expression of lytM, a unique autolytic gene of Staphylococcus aureus. It has also been used in a study to investigate the sequence analysis of a Staphylococcus aureus gene encoding a peptidoglycan hydrolase activity.

Biochem/physiol Actions

A protein complex with highly specific lytic activity against Staphylococcus species, including Staphylococcus aureus.
Lysostaphin possesses a lytic action against Staphylococcus aureus, which is used to treat antibiotic-resistant staphylococcal infections. It has the properties of enzymes, such as glycylglycine endopeptidase, endo-β-N-acetyl glucosamidase and N-acetyl muramyl-L-alanine amidase. Lysostaphin is also used as a preservative in food industry and in clinical labs for rapid screening.
The antimicrobial activity of lysostaphin, a metalloendopeptidase expressed by Staphylococcus simulans, is enhanced by binding to the C′ terminus of the membrane-associated Trap protein of Staphylococcus aureus.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X Wang et al.
Gene, 102(1), 105-109 (1991-06-15)
The nucleotide (nt) sequence of a 2.0-kb NheI-XbaI DNA fragment containing a peptidoglycan hydrolase-encoding gene, lytA, tentatively identified as encoding an N-acetylmuramyl-L-alanine amidase, from Staphylococcus aureus, was determined. The nt sequencing revealed an open reading frame (ORF) of 1443 bp
C-terminus of TRAP in Staphylococcus can enhance the activity of lysozyme and lysostaphin.
Yang, G.
Acta Biochimica et Biophysica Hungarica, 40, 452-458 (2008)
L Ramadurai et al.
Journal of bacteriology, 179(11), 3625-3631 (1997-06-01)
A gene encoding an autolytic activity was identified in an autolysis-deficient mutant (Lyt-) of Staphylococcus aureus which produces only a single band in autolytic-activity gels (N. Mani, P. Tobin, and R. K. Jayaswal, J. Bacteriol. 175:1493-1499, 1993). An open reading
Lysostaphin: an antistaphylococcal agent
Kumar J K
Applied Microbiology and Biotechnology, 80(4), 555-561 (2008)
Grégory Francius et al.
Journal of bacteriology, 190(24), 7904-7909 (2008-10-07)
The advent of Staphylococcus aureus strains that are resistant to virtually all antibiotics has increased the need for new antistaphylococcal agents. An example of such a potential therapeutic is lysostaphin, an enzyme that specifically cleaves the S. aureus peptidoglycan, thereby

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service