Merck
  • BK Virus replication in vitro: limited effect of drugs interfering with viral uptake and intracellular transport.
BK Virus replication in vitro: limited effect of drugs interfering with viral uptake and intracellular transport.
Antimicrobial agents and chemotherapy (2007-09-26)
Parmjeet Randhawa, Noush Afarin Farasati, Yuchen Huang
ABSTRACT
BK virus is an important pathogen in kidney transplant recipients. In vitro studies demonstrated slight antiviral activity for chloroquine and nystatin. A sialic acid derivative, BTB11968, was identified as a lead compound for further development.
Sigma-Aldrich
Octyl β-D-glucopyranoside, BioXtra, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside, ≥98% (GC)
Sigma-Aldrich
Octyl β-D-glucopyranoside solution, ≥95% (HPLC), 50 % (w/v) in H2O
Sigma-Aldrich
Nonyl β-D-glucopyranoside, ≥97.0% (GC)
Sigma-Aldrich
Colchicine, BioReagent, suitable for plant cell culture, ≥95% (HPLC)
Sigma-Aldrich
Methyl α-D-mannopyranoside, ≥99.0% (HPLC)
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Millipore
Methyl α-D-mannopyranoside, ≥99.0%, suitable for microbiology, enables differentiation between species of Listeria