Merck
  • Structure-activity relationships of 1'S-1'-acetoxychavicol acetate for inhibitory effect on NO production in lipopolysaccharide-activated mouse peritoneal macrophages.
Structure-activity relationships of 1'S-1'-acetoxychavicol acetate for inhibitory effect on NO production in lipopolysaccharide-activated mouse peritoneal macrophages.
Bioorganic & medicinal chemistry letters (2005-03-23)
Hisashi Matsuda, Shin Ando, Toshio Morikawa, Shinya Kataoka, Masayuki Yoshikawa
ABSTRACT
1'S-1'-Acetoxychavicol acetate from the rhizomes of Alpinia galanga inhibited nitric oxide (NO) production in lipopolysaccharide-activated mouse peritoneal macrophages with an IC(50) value of 2.3 microM. To clarify the structure-activity relationship of 1'S-1'-acetoxychavicol acetate, various natural and synthetic phenylpropanoids and synthetic phenylbutanoids were examined, and the following structural requirements were clarified. (1) The para or ortho substitution of the acetoxyl and 1-acetoxypropenyl groups at the benzene ring was essential. (2) The S configuration of the 1'-acetoxyl group was preferable. (3) The presence of the 3-methoxyl group and disappearance of the 2'-3' double bond by hydrogenation reduced the activity. (4) The substitution of acetyl groups with propionyl or methyl groups reduced the activity. (5) Lengthening of the carbon chain between the 1'- and 2'-positions reduced the activity.
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Sigma-Aldrich
4-Allyl-1,2-dimethoxybenzene, 99%
Sigma-Aldrich
trans-3-Hydroxycinnamic acid, 99%
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Sigma-Aldrich
Acetone, ≥99%, meets FCC analytical specifications
Supelco
Acetone, analytical standard
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
(S)-(−)-1-Phenyl-1-propanol, 99%
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Methyl eugenol, ≥98%, FCC
Sigma-Aldrich
Sinapic acid, ≥98%, powder
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.0% (T)
Supelco
Sinapic acid, matrix substance for MALDI-MS, ≥99.5%, Ultra pure
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
Sigma-Aldrich
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.8%
Sigma-Aldrich
trans-Cinnamic acid, 97%
Sigma-Aldrich
4-Methoxycinnamic acid, predominantly trans, 99%
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Supelco
Caffeic acid, matrix substance for MALDI-MS, ≥99.0% (HPLC)
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)