Merck
  • Enhanced neprilysin-mediated degradation of hippocampal Aβ42 with a somatostatin peptide that enters the brain.
Enhanced neprilysin-mediated degradation of hippocampal Aβ42 with a somatostatin peptide that enters the brain.
Theranostics (2021-01-05)
Fadi Rofo, Canan Ugur Yilmaz, Nicole Metzendorf, Tobias Gustavsson, Chiara Beretta, Anna Erlandsson, Dag Sehlin, Stina Syvänen, Per Nilsson, Greta Hultqvist
ABSTRACT
Background: Aggregation of the amyloid-beta (Aβ) peptide is one of the main neuropathological events in Alzheimer's disease (AD). Neprilysin is the major enzyme degrading Aβ, with its activity enhanced by the neuropeptide somatostatin (SST). SST levels are decreased in the brains of AD patients. The poor delivery of SST over the blood-brain barrier (BBB) and its extremely short half-life of only 3 min limit its therapeutic significance. Methods: We recombinantly fused SST to a BBB transporter binding to the transferrin receptor. Using primary neuronal cultures and neuroblastoma cell lines, the ability of the formed fusion protein to activate neprilysin was studied. SST-scFv8D3 was administered to mice overexpressing the Aβ-precursor protein (AβPP) with the Swedish mutation (APPswe) as a single injection or as a course of three injections over a 72 h period. Levels of neprilysin and Aβ were quantified using an Enzyme-linked immunosorbent assay (ELISA). Distribution of SST-scFv8D3 in the brain, blood and peripheral organs was studied by radiolabeling with iodine-125. Results: The construct, SST-scFv8D3, exhibited 120 times longer half-life than SST alone, reached the brain in high amounts when injected intravenously and significantly increased the brain concentration of neprilysin in APPswe mice. A significant decrease in the levels of membrane-bound Aβ42 was detected in the hippocampus and the adjacent cortical area after only three injections. Conclusion: With intravenous injections of our BBB permeable SST peptide, we were able to significantly increase the levels neprilysin, an effect that was followed by a significant and selective degradation of membrane-bound Aβ42 in the hippocampus. Being that membrane-bound Aβ triggers neuronal toxicity and the hippocampus is the central brain area in the progression of AD, the study has illuminated a new potential treatment paradigm with a promising safety profile targeting only the disease affected areas.
Sigma-Aldrich
N-Succinyl-Ala-Ala-Phe-7-amido-4-methylcoumarin, protease substrate
Sigma-Aldrich
Leucine Aminopeptidase, microsomal from porcine kidney, Type IV-S, ammonium sulfate suspension, 10-40 units/mg protein (Bradford)
Sigma-Aldrich
N-Acetyl-D-penicillamine, for HPLC derivatization, ≥99.0% (T)
Sigma-Aldrich
DL-Thiorphan, ≥98% (TLC)
Sigma-Aldrich
Goat Anti-Mouse IgG Antibody, HRP conjugate, Upstate®, from goat
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
Chloramine-T hydrate, 95%
Sigma-Aldrich
Valproic acid sodium salt, 98%
Sigma-Aldrich
Goat Anti-Rat IgG Antibody, HRP conjugate, Chemicon®, from goat
Sigma-Aldrich
Anti-Somatostatin Antibody, clone YC7, culture supernatant, clone YC7, Chemicon®