Skip to Content
Merck
All Photos(5)

Key Documents

MAB354

Sigma-Aldrich

Anti-Somatostatin Antibody, clone YC7

culture supernatant, clone YC7, Chemicon®

Synonym(s):

Anti-Somatostatin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rat

Quality Level

antibody form

culture supernatant

antibody product type

primary antibodies

clone

YC7, monoclonal

species reactivity

rabbit, rat, human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SST(6750)

General description

Somatostatin (also known as growth hormone inhibiting hormone (GHIH) or somatotropin release-inhibiting factor (SRIF) has two active forms, 14 amino acids and 28 amino acids, which are produced by alternate cleavage of the single preproprotein. Somatostatin is expressed widely and acts to inhibit the release of several secondary hormones by binding to high-affinity G-protein coupled somatostatin receptors. Presence of somatostatin has been shown in sympathetic nerves, mucosal cells, myenteric nerves of the gastrointestinal tract, salivary glands and in some parafollicular cells of the thyroid.

Specificity

Anti-SST, clone YC7 recognizes Somatostatin. Shows no cross-reactivity to enkephalins, other endorphins, substance P or CGRP. Partially cross-reacts with somatostatin fragments.

Immunogen

Synthetic peptide corresponding to amino acids 1-14 of cyclic somatostatin conjugated to bovine thyroglobulin using carbodiimide.

Application

Anti-Somatostatin Antibody, clone YC7 is an antibody against Somatostatin for use in IH.
Immunohistochemistry at 1:50-1:100 on human brain.

Optimal working dilutions must be determined by the end user.

Physical form

Lyophilized Supernatant, preservative free.

Storage and Stability

Lyophilized antibody can be kept at 2-8°C for up to 3 months and should be kept at -20°C for long term storage. Reconstitute with 100μL sterile DI water. Maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Analysis Note

Control
Pancreas, Brain

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recruitment of Perisomatic Inhibition during Spontaneous Hippocampal Activity In Vitro.
Beyeler, A; Retailleau, A; Molter, C; Mehidi, A; Szabadics, J; Leinekugel, X
Testing null
Cell type-specific inhibitory inputs to dendritic and somatic compartments of parvalbumin-expressing neocortical interneuron.
Hioki, H; Okamoto, S; Konno, M; Kameda, H; Sohn, J; Kuramoto, E; Fujiyama, F; Kaneko, T
The Journal of Neuroscience null
Csaba Varga et al.
Cerebral cortex (New York, N.Y. : 1991), 25(11), 4430-4449 (2015-03-20)
Transcription factors contribute to the differentiation of cortical neurons, orchestrate specific interneuronal circuits, and define synaptic relationships. We have investigated neurons expressing chicken ovalbumin upstream promoter transcription factor II (COUP-TFII), which plays a role in the migration of GABAergic neurons.
Txomin Lalanne et al.
The Journal of physiology, 594(4), 837-861 (2015-11-06)
In the hippocampus, calcium-permeable AMPA receptors have been found in a restricted subset of neuronal types that inhibit other neurons, although their localization in the neocortex is less well understood. In the present study, we looked for calcium-permeable AMPA receptors
Latefa Yekhlef et al.
Journal of neurophysiology, 113(5), 1616-1630 (2014-12-17)
GABAergic interneurons are thought to play a critical role in eliciting interictal spikes (IICs) and triggering ictal discharges in temporal lobe epilepsy, yet the contribution of different interneuronal subtypes to seizure initiation is still largely unknown. Here we took advantage

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service