Merck
  • DYRK kinase Pom1 drives F-BAR protein Cdc15 from the membrane to promote medial division.
DYRK kinase Pom1 drives F-BAR protein Cdc15 from the membrane to promote medial division.
Molecular biology of the cell (2020-02-27)
Rahul Bhattacharjee, MariaSanta C Mangione, Marcin Wos, Jun-Song Chen, Chloe E Snider, Rachel H Roberts-Galbraith, Nathan A McDonald, Libera Lo Presti, Sophie G Martin, Kathleen L Gould
ABSTRACT
In many organisms, positive and negative signals cooperate to position the division site for cytokinesis. In the rod-shaped fission yeast Schizosaccharomyces pombe, symmetric division is achieved through anillin/Mid1-dependent positive cues released from the central nucleus and negative signals from the DYRK-family polarity kinase Pom1 at cell tips. Here we establish that Pom1's kinase activity prevents septation at cell tips even if Mid1 is absent or mislocalized. We also find that Pom1 phosphorylation of F-BAR protein Cdc15, a major scaffold of the division apparatus, disrupts Cdc15's ability to bind membranes and paxillin, Pxl1, thereby inhibiting Cdc15's function in cytokinesis. A Cdc15 mutant carrying phosphomimetic versions of Pom1 sites or deletion of Cdc15 binding partners suppresses division at cell tips in cells lacking both Mid1 and Pom1 signals. Thus, inhibition of Cdc15-scaffolded septum formation at cell poles is a key Pom1 mechanism that ensures medial division.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Brain Extract from bovine brain, Type I, Folch Fraction I
Sigma-Aldrich
Monoclonal Anti-PSTAIR antibody produced in mouse, clone PSTAIR, ascites fluid
Sigma-Aldrich
2-Mercaptoethanol, for molecular biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
Calcofluor White Stain, suitable for microbiology
Sigma-Aldrich
3X FLAG® Peptide, lyophilized powder
Millipore
Anti-FLAG® M2 Magnetic Beads, affinity isolated antibody
Sigma-Aldrich
Benzamidine, ≥95.0%
Sigma-Aldrich
Maltose solution, for molecular biology, BioReagent, ~20% in H2O
Sigma-Aldrich
Brilliant Blue G, pure
Millipore
ANTI-FLAG® High Sensitivity, M2 coated 96-well plates, 96-well, clear, polystyrene, flat bottom plate
Sigma-Aldrich
Benzamidine hydrochloride hydrate, ≥99%
Sigma-Aldrich
Methyl Blue, powder or crystals
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Lysozyme from chicken egg white, powder or granules, ≥90 %, ≥39,000 units/mg protein
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Millipore
GST•Bind Resin