Merck
  • Evaluation of multi-starter S. cerevisiae/ D. bruxellensis cultures for mimicking and accelerating transformations occurring during barrel ageing of beer.
Evaluation of multi-starter S. cerevisiae/ D. bruxellensis cultures for mimicking and accelerating transformations occurring during barrel ageing of beer.
Food chemistry (2020-04-27)
Eduardo Coelho, Mário Azevedo, José A Teixeira, Teresa Tavares, José M Oliveira, Lucília Domingues
ABSTRACT
During beer ageing, endogenous barrel microbes grow spontaneously and transform wort/beer composition, being Dekkera bruxellensis and Saccharomyces cerevisiae among the main contributors to the chemical and sensory profile of aged beer. This work aims at the application of multi-starter cultures to mimic and accelerate biological modifications occurring during barrel ageing of beer, in controlled fermentation processes. Co-cultures of D.bruxellensis/S.cerevisiae were conducted under conditions commonly found in barrel aged beer production: different pitching rates, high glucose concentration and presence of ethanol and wood extracts. Selective pressures and competition between yeasts influenced microbial growth and metabolite production, namely ethanol, acetic acid and target volatile compounds (esters, alcohols, terpenols, volatile acids and volatile phenols). Metabolic profiles of co-cultures combined traits of both species, and differed from those of pure cultures. Lastly, multi-starters were successfully applied in combination with wood in a controlled and accelerated fermentation process for mimicking barrel ageing transformations.
Sigma-Aldrich
(±)-2-Methyl-1-butanol, ≥98.0% (GC)
Sigma-Aldrich
Ethyl hydrocinnamate, 99%
Sigma-Aldrich
Ethyl decanoate, ReagentPlus®, ≥99%
Sigma-Aldrich
Ethyl phenylacetate, ReagentPlus®, 99%
Sigma-Aldrich
Isobutyric acid, puriss. p.a., ≥99.5%
Sigma-Aldrich
Diethyl malate, ≥97%, FG
Sigma-Aldrich
Isopentyl acetate, anhydrous, ≥99%
Sigma-Aldrich
4-Ethylguaiacol, ≥98%, FCC, FG
Sigma-Aldrich
Geraniol, 98%
Sigma-Aldrich
Ethyl octanoate, ReagentPlus®, ≥99%
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Ethyl butyrate, 99%
Sigma-Aldrich
Ethyl hexanoate, ≥99%
Sigma-Aldrich
5-Methylfurfural, ReagentPlus®, 99%
Sigma-Aldrich
2-(4-Hydroxyphenyl)ethanol, 98%
Sigma-Aldrich
4-Ethylphenol, 99%