112321
Ethyl octanoate
ReagentPlus®, ≥99%
Synonym(s):
Ethyl caprylate
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Linear Formula:
CH3(CH2)6COOC2H5
CAS Number:
Molecular Weight:
172.26
Beilstein:
1754470
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
vapor pressure
0.02 mmHg ( 25 °C)
Quality Level
product line
ReagentPlus®
Selectophore™
Assay
≥99%
refractive index
n20/D 1.417 (lit.)
bp
206-208 °C (lit.)
mp
−48-−47 °C (lit.)
density
0.867 g/mL at 20 °C (lit.)
functional group
ester
SMILES string
CCCCCCCC(=O)OCC
InChI
1S/C10H20O2/c1-3-5-6-7-8-9-10(11)12-4-2/h3-9H2,1-2H3
InChI key
YYZUSRORWSJGET-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Ethyl octanoate is a powerful odourant found in mead (a traditional drink that contains alcohol).
Application
Ethyl octanoate, found in Burgundy wine, was used to study its partition co-efficient in water in model wine using the headspace method.
Legal Information
ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany
Selectophore is a trademark of Merck KGaA, Darmstadt, Germany
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
167.0 °F - closed cup
Flash Point(C)
75 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Influence of Yeast Walls on the Behavior of Aroma Compounds in a Model Wine.
Lubbers S, et al.
American Journal of Enology and Viticulture, 45(1), 29-33 (1994)
A P Pereira et al.
Food microbiology, 33(1), 114-123 (2012-11-06)
Mead is a traditional drink that contains 8%-18% (v/v) of ethanol, resulting from the alcoholic fermentation of diluted honey by yeasts. Mead fermentation is a time-consuming process and the quality of the final product is highly variable. Therefore, the present
Eduardo Coelho et al.
Molecules (Basel, Switzerland), 25(14) (2020-07-19)
Cassava plays a key role in the food production and economies of several countries worldwide. Due to its starch content, alcoholic fermentation is a promising transformation process for adding value to cassava. However, most of the existing cassava beverages are
Tanya E Rutan et al.
Journal of agricultural and food chemistry, 66(38), 10053-10066 (2018-09-04)
The effects of two cluster thinning regimes (low and moderate) with Vitis vinifera cv. Pinot noir, in vineyards located in Central Otago, New Zealand, on wine composition, were studied across three consecutive seasons. There were strong correlations between the extent
Zuobing Xiao et al.
Journal of agricultural and food chemistry, 67(32), 8926-8937 (2019-07-30)
The volatile compounds found in mangoes (Mangifera indica L.) obtained from three cultivars (TN, Tainong no. 1; KT, Keitt; and ZL, Zill; grown in China) were comprehensively analyzed by gas chromatography-olfactometry (GC-O), gas chromatography-mass spectrometry (GC-MS), and GC-flame photometric detection
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service