Merck
  • Pbrm1 Steers Mesenchymal Stromal Cell Osteolineage Differentiation by Integrating PBAF-Dependent Chromatin Remodeling and BMP/TGF-β Signaling.
Pbrm1 Steers Mesenchymal Stromal Cell Osteolineage Differentiation by Integrating PBAF-Dependent Chromatin Remodeling and BMP/TGF-β Signaling.
Cell reports (2020-04-30)
Sayantani Sinha, Mayukh Biswas, Shankha Subhra Chatterjee, Sanjay Kumar, Amitava Sengupta
ABSTRACT
Bone morphogenic protein (BMP)/transforming growth factor β (TGF-β) signaling determines mesenchymal-stromal-cell (MSC) osteolineage commitment and tissue identity. However, molecular integration of developmental signaling with MSC-intrinsic chromatin regulation remains incompletely understood. SWI/SNF-(BAF) is an ATP-dependent chromatin remodeler implicated in multi-cellular development. We show that BMPs and long-term osteogenic signals in MSCs selectively induce expression of polybromo BAF (PBAF) components Pbrm1, Arid2, and Brd7. Loss of Pbrm1/Arid2/Brd7 profoundly impairs osteolineage gene expression and osteogenesis without compromising adipogenesis. Pbrm1 loss attenuates MSC in vivo ossification. Mechanistically, Pbrm1/PBAF deficiency impairs Smad1/5/8 activation through locus-specific epi-genomic remodeling, involving Pbrm1 bromodomains, along with transcriptional downregulation of Bmpr/TgfβrII affecting BMP-early-responsive gene expression. Gain of function of BmprIβ, TgfβrII in PBAF-deficient MSCs partly restores Smad1/5/8 activation and osteogenesis. Pbrm1 loss further affects hematopoietic stem and progenitor activity through non-cell-autonomous regulation of microenvironment and niche-factor expression. Together, these findings reveal a link illustrating epi-genomic feedforward control of BMP/TGF-β signaling to transcriptional and cellular plasticity in the mesenchymal microenvironment and account for stromal-SWI/SNF in hematopoiesis.
Sigma-Aldrich
Indomethacin, 98.5-100.5% (in accordance with EP)
Sigma-Aldrich
7-Aminoactinomycin D, ~97% (HPLC), powder
Sigma-Aldrich
Ribonuclease A from bovine pancreas, for molecular biology, ≥70 Kunitz units/mg protein, lyophilized
Sigma-Aldrich
Insulin human, recombinant, expressed in yeast (proprietary host)
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Anti-BAF250a/ARID1a Antibody, clone PSG3, clone PSG3, Upstate®, from mouse
Sigma-Aldrich
Anti-SNF2β/BRG1 Antibody, serum, Upstate®
Supelco
GR-2 Septa, diam. 11 mm ( 7/16 in.)
Sigma-Aldrich
β-Glycerol phosphate disodium salt pentahydrate, ≥98.0% (NT)
Sigma-Aldrich
Alizarin Red S, certified by the Biological Stain Commission
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
SIGMAFAST BCIP®/NBT, tablet
Sigma-Aldrich
Proteinase K from Tritirachium album, lyophilized powder, BioUltra, ≥30 units/mg protein, for molecular biology