Skip to Content
Merck
All Photos(1)

Key Documents

B5655

Sigma-Aldrich

BCIP®/NBT

SIGMAFAST, alkaline phosphatase substrate, chromogenic, tablet

Synonym(s):

BCIP/NBT Substrate, BCIP®/NBT Alkaline Phosphatase Substrate

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.83

product name

SIGMAFAST BCIP®/NBT, tablet

form

tablet

Quality Level

solubility

water: 10 mL, clear, yellow

storage temp.

−20°C

Application

SIGMAFAST BCIP®/NBT has been used as a substrate
  • for the anti-mouse IgG conjugated alkaline phosphatase in immunoblotting
  • for alkaline phosphatase in the histochemical staining of primary osteoblasts
  • for alkaline phosphatase staining in bone marrow stromal cells

Reconstitution

Each tablet dissolved in 10 ml deionized water yields a ready-to-use buffered solution containing BCIP/NBT, pH 9.5.

Legal Information

BCIP is a registered trademark of Merck KGaA, Darmstadt, Germany
SIGMAFAST is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sialic acid on herpes simplex virus type 1 envelope glycoproteins is required for efficient infection of cells
Teuton JR and Brandt CR
Journal of Virology, 81(8), 3731-3739 (2007)
Nicolas Leventoux et al.
Stem cell research, 47, 101896-101896 (2020-07-14)
Amyotrophic Lateral Sclerosis is the most common motor neuron degenerative disease in adults, and TARDBP gene mutations have been reported to be involved in the pathogenesis. We present here how we generated the human induced pluripotent stem cell (hiPSC) line
Mitogen-activated protein kinase phosphatase 1 regulates bone mass, osteoblast gene expression, and responsiveness to parathyroid hormone
Mahalingam CD, et al.
The Journal of Endocrinology, 211(2), 145-156 (2011)
Aneela Yasmeen et al.
Molecular biotechnology, 58(12), 807-820 (2016-10-21)
The conserved coat or V2 gene of begomoviruses is responsible for viral movement in the plant cells. RNAi technology was used to silence V2 gene for resistance against these viruses in transgenic plants. The transformation of the RNAi-based gene construct
MicroRNA-183-5p increases with age in bone-derived extracellular vesicles, suppresses bone marrow stromal (stem) cell proliferation, and induces stem cell senescence
Davis C, et al.
Tissue Engineering: Part A, 23(21-22), 1231-1240 (2017)

Protocols

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

BCIP/NBT substrate produces stable, intense color for alkaline phosphatase assays, ideal for visual observation.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service