Skip to Content
Merck
All Photos(3)

Key Documents

R6513

Sigma-Aldrich

Ribonuclease A from bovine pancreas

for molecular biology, ≥70 Kunitz units/mg protein, lyophilized

Synonym(s):

Pancreatic Ribonuclease, RNAsea, RNase A, Ribonucleate 3′-pyrimidinooligonucleotidohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.53

grade

for molecular biology

Quality Level

form

lyophilized

specific activity

≥70 Kunitz units/mg protein

mol wt

13.7 kDa
~13,700

foreign activity

Exonuclease and endonuclease, none detected
NICKase and DNase, none detected

storage temp.

−20°C

SMILES string

[nH]1cnc(c1)CC(NC(=O)CCN)C(=O)O

InChI

1S/C9H14N4O3/c10-2-1-8(14)13-7(9(15)16)3-6-4-11-5-12-6/h4-5,7H,1-3,10H2,(H,11,12)(H,13,14)(H,15,16)

InChI key

CQOVPNPJLQNMDC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

RNase A is a single chain polypeptide containing 124 amino acids, linked by four disulfide bridges. In contrast to RNase B, RNase A is not a glycoprotein. RNase A can be inhibited by alkylation of His12 or His119, which are present in the active site of the enzyme. Activators of RNase A include potassium and sodium salts.
RNase A is an endoribonuclease that attacks at the 3′OHphosphate of a pyrimidine nucleotide. The sequence of pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG. The highest activity is exhibited with single stranded RNA.
RNase A, Ribonuclease A, is an endoribonuclease that cleaves the phosphodiester bonds of single strand RNA after pyrimidine nucleotides. It attacks at the 3′ phosphate end (For example pG-pG-pC-pA-pG will be cleaved to give pG-pG-pCp and A-pG). The highest activity is exhibited with single stranded RNA. RNase A is a single chain polypeptide containing 4 disulfide bridges. In contrast to RNase B, it is not a glycoprotein. Ribonucleases do not hydrolyze DNA, because the DNA lacks 2′-OH groups essential for the formation of cyclic intermediates. RNase A can also hydrolyze RNA from protein samples. RNase A can be inhibited by alkylation of His12 and His119 and activated by potassium and sodium salts. RNAse is inhibited in the presence of heavy metal ions. RNase is also inhibited competitively by DNA.

Application

  • RNase A is used to remove RNA from DNA plasmid and genomic DNA preparations and protein samples.
  • RNase A is also used in RNA sequence analysis and protection assays.
  • RNase A has been used as a tool for computer-aided drug design.
  • RNase A supports the analysis of RNA sequences.
  • RNase A hydrolyze RNA contained in protein samples.
  • Purification of DNA is supported by RNase A.
Ribonuclease A from bovine pancreas has been used:
  • for plasmid DNA purification
  • to digest and avoid staining of double-stranded RNA during staining of nuclei
  • in immunoperoxidase and immunofluorescence detection
  • to treat cells prior to flow cytometry
  • as a component of post-hybridization buffer for washing FFPE tissue sections post in-situ hybridization
  • to treat cells during the preparation of outer membrane protein (OMP) extracts
  • RNase protection assays
  • Removal of unspecifically bound RNA
  • Analysis of RNA sequences
  • Hydrolysis of RNA contained in protein samples

Features and Benefits

Our highly stable Ribonuclease A, RNase A, is suitable for removal of RNA, RNA sequencing, and DNA purification.

Analysis Note

Protein determined by E.

Other Notes

A major application for RNase A is the removal of RNA from preparations of plasmid DNA. For this application, DNase free RNase A is used at a final concentration of 10 μg/mL.

Boiling stock solutions of this RNase A product to inactivate residual DNase is not necessary and may cause precipitation of RNase and possible loss of enzymatic activity. If an RNase A solution is heated at a neutral pH, precipitation will occur. When heated at a lower pH, some precipitation may occur because of protein impurities that are present.
Activators of RNase A include potassium and sodium salts. RNase A can be inhibited by alkylation of His12 or His119.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santella, Regina M., and Yu-Jing Zhang
DNA Fingerprinting: An Introduction (2011)
Kento Kurata et al.
International journal of oncology, 49(6), 2303-2308 (2016-10-18)
Anaplastic thyroid cancer (ATC) is a rare malignancy that progresses extremely aggressively and often results in dismal prognosis. We investigated the efficacy of inhibiting the activated RAS/RAF/MEK pathway in ATC cells aiming to clarify the mechanism of effect and resistance.
Production of hen egg yolk immunoglobulins simultaneously directed against Salmonella Enteritidis and Salmonella Typhimurium in the same egg yolk
Chalghoumi, Raja, et al.
Poultry Science (2008)
Rapid and irreversible CD4+ T-cell depletion induced by the highly pathogenic simian/human immunodeficiency virus SHIVDH12R is systemic and synchronous
Igarashi Tatsuhiko, et al.
Journal of Virology (2002)
Berg JM, Tymoczko JL, Stryer L.
BioChemistry: An Indian Journal (2002)

Protocols

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

This procedure may be used for determination of Ribonuclease A (RNase A) activity.

Related Content

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

KOD One™ PCR Master Mix overview for ultra-fast PCR with high specificity, fidelity, and yield

Chromatograms

application for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service