Merck
  • The RGD motif is involved in CD97/ADGRE5-promoted cell adhesion and viability of HT1080 cells.
The RGD motif is involved in CD97/ADGRE5-promoted cell adhesion and viability of HT1080 cells.
Scientific reports (2019-02-08)
Wen-Ye Tjong, Hsi-Hsien Lin
ABSTRACT
CD97/ADGRE5 is an adhesion G protein-coupled receptor (aGPCR) involved in tumor cell adhesion, migration, angiogenesis, and apoptosis. CD97 has been shown previously to stimulate angiogenesis by interacting with integrins on endothelial cells via an Arginine-Glycine-Aspartic acid (RGD) motif. In this report, the role of the RGD motif in tumor cell adhesion and apoptosis was investigated using a previously-established HT1080 cell-based system. We found that the RGD motif is critical in CD97-promoted cell adhesion, in part due to the up-regulation of αvβ5 and α2β1 integrins, and that CD97 mediates its anti-apoptotic effect in extrinsic apoptosis via RGD-dependent cell adhesion. In contrast, CD97-modulated anti-apoptotic effect in intrinsic apoptosis is mediated by RGD-independent, N-cadherin-induced homotypic cell aggregation. Hence, CD97 promotes tumorigenesis via RGD-dependent and -independent mechanisms.
Sigma-Aldrich
Anti-Integrin αVβ5 Antibody, clone P1F6, azide free, clone P1F6, Chemicon®, from mouse
Sigma-Aldrich
Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62, clone ab62, from mouse
Sigma-Aldrich
Anti-Integrin β1 Antibody, activated, clone HUTS-4, Azide Free, clone HUTS-4, Chemicon®, from mouse
Sigma-Aldrich
Anti-Integrin α2β1 Antibody, clone BHA2.1, azide free, clone BHA2.1, Chemicon®, from mouse