Skip to Content
Merck
All Photos(1)

Key Documents

MABT27

Sigma-Aldrich

Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62

clone ab62, from mouse

Synonym(s):

LIBS2 domain, Integrin, Integrin alphaVbeta3, LIBS2 epitiope

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ab62, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
western blot: suitable

isotype

IgG2aκ

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ITGB3(3690)

General description

The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The involvement of integrins in vascular proliferation, adhesion, and wound repair has been well documented. The adhesion receptor, integrin alphaVbeta3, appears to be selectively expressed on growing blood vessels and has been identified as a marker of angiogenic vascular tissue.
Clone Ab62 is a monoclonal antibody that specifically recognizes the LIBS2 epitope in the C-terminal region of the extracellular domain of the beta3 subunit (Wiberzbicka-Patynowski, I., et al. (1999). JBC. 274(53):37809-37814).

Specificity

This antibody recognizes the LIBS2 epitope in the C-terminal region of the extracellular domain of the beta3 subunit of integrins.

Immunogen

Epitope: LIBS2 epitope in the C-terminal region of the extracellular domain of the beta3 subunit of integrins.
Human GPIIa-IIIb purified from platelets

Application

Anti-LIBS2 epitope (Integrin beta3 subunit) Antibody, clone ab62 is an antibody against LIBS2 epitope (Integrin beta3 subunit) for use in WB, FC.
Flow Cytometry Analysis: A previous lot was used by an independent laboratory in FC (Ginsberg, M., et al. (1990). Blood. 76:2017-2023.)
Research Category
Cell Structure
Research Sub Category
Integrins

Quality

Evaluated by Western Blot in human platelets cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected LIBS2 domain of integrin beta3 subunit in 10 µg of human platelets cell lysate.

Target description

~110 kDa observed in human platelets.
Other MW bands may vary or may be observed depending on lysate used and if the LIBS2 epitope is present in other proteins.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human platelets cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhen Feng et al.
Proceedings of the National Academy of Sciences of the United States of America, 118(19) (2021-05-06)
Invadopodia are integrin-mediated adhesions with abundant PI(3,4)P2 However, the functional role of PI(3,4)P2 in adhesion signaling remains unclear. Here, we find that the PI(3,4)P2 biogenesis regulates the integrin endocytosis at invadopodia. PI(3,4)P2 is locally produced by PIK3CA and SHIP2 and
Rubén A Bartolomé et al.
Molecular oncology, 15(7), 1849-1865 (2021-03-15)
Cadherin 6 (CDH6) is significantly overexpressed in advanced ovarian and renal cancers. However, the role of CDH6 in cancer metastasis is largely unclear. Here, we investigated the impact of CDH6 expression on integrin-mediated metastatic progression. CDH6 preferentially bound to αIIbβ3
F Zhou et al.
Lab on a chip, 18(19), 2966-2974 (2018-09-01)
Cell adhesion plays a critical role in many cellular functions, such as the hemostatic/thrombotic process, inflammatory reactions, and adaptive immune response. Many cell adhesion processes involve crosstalk between multiple ligand-receptor systems through intracellular signaling. To elucidate such crosstalk requires analysis
Wen-Ye Tjong et al.
Scientific reports, 9(1), 1517-1517 (2019-02-08)
CD97/ADGRE5 is an adhesion G protein-coupled receptor (aGPCR) involved in tumor cell adhesion, migration, angiogenesis, and apoptosis. CD97 has been shown previously to stimulate angiogenesis by interacting with integrins on endothelial cells via an Arginine-Glycine-Aspartic acid (RGD) motif. In this
Anna Testa et al.
Matrix biology : journal of the International Society for Matrix Biology, 124, 49-62 (2023-11-14)
Highly aggressive, metastatic, neuroendocrine prostate cancer, which typically develops from prostate cancer cells acquiring resistance to androgen deprivation therapy, is associated with limited treatment options and hence poor prognosis. We have previously demonstrated that the αVβ3 integrin is over-expressed in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service