Merck
  • Mechanically stimulated ATP release from murine bone cells is regulated by a balance of injury and repair.
Mechanically stimulated ATP release from murine bone cells is regulated by a balance of injury and repair.
eLife (2018-10-17)
Nicholas Mikolajewicz, Elizabeth A Zimmermann, Bettina M Willie, Svetlana V Komarova
ABSTRACT
Bone cells sense and actively adapt to physical perturbations to prevent critical damage. ATP release is among the earliest cellular responses to mechanical stimulation. Mechanical stimulation of a single murine osteoblast led to the release of 70 ± 24 amole ATP, which stimulated calcium responses in neighboring cells. Osteoblasts contained ATP-rich vesicles that were released upon mechanical stimulation. Surprisingly, interventions that promoted vesicular release reduced ATP release, while inhibitors of vesicular release potentiated ATP release. Searching for an alternative ATP release route, we found that mechanical stresses induced reversible cell membrane injury in vitro and in vivo. Ca2+/PLC/PKC-dependent vesicular exocytosis facilitated membrane repair, thereby minimizing cell injury and reducing ATP release. Priming cellular repair machinery prior to mechanical stimulation reduced subsequent membrane injury and ATP release, linking cellular mechanosensitivity to prior mechanical exposure. Thus, our findings position ATP release as an integrated readout of membrane injury and repair.
Sigma-Aldrich
Quinacrine dihydrochloride, ≥90%
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Carbenoxolone disodium salt, ≥98%
Sigma-Aldrich
Adenosine 5′-triphosphate magnesium salt, ≥95%, bacterial
Sigma-Aldrich
ML218, ≥98% (HPLC)
Sigma-Aldrich
GSK2193874, ≥98% (HPLC)
Sigma-Aldrich
Venor GeM Mycoplasma Detection Kit, PCR-based
Sigma-Aldrich
Bisindolylmaleimide II, ≥97% (Mixture of 2 isomers)
Sigma-Aldrich
Luciferase from Photinus pyralis (firefly), recombinant, expressed in E. coli, buffered aqueous solution, ≥10×1010 units/mg protein