Skip to Content
Merck
All Photos(1)

Key Documents

MP0025

Sigma-Aldrich

Venor GeM Mycoplasma Detection Kit, PCR-based

Synonym(s):

PCR mycoplasma detection kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.75
Pricing and availability is not currently available.

usage

 kit sufficient for 25 tests

Quality Level

packaging

pkg of 1 kit

storage condition

dry at room temperature

technique(s)

PCR: suitable

application(s)

detection
microbiology

storage temp.

2-8°C

General description

The Venor GeM Mycoplasma Detection Kit utilizes the polymerase chain reaction (PCR), which was established as the method of choice for highest sensitivity in the detection of Mycoplasma and Acholeplasma contamination in cell cultures and other cell culture derived biologicals. Detection requires as little as 1–5fg of mycoplasma DNA corresponding to 2–5 mycoplasma per sample volume. The primer set is specific to the highly conserved rRNA operon, or more specifically, the 16S rRNA coding region in the mycoplasma genome. This allows for detection of all Mycoplasma, Acholeplasma and Ureaplasma species tested so far, which are usually encountered as contaminants in cell cultures. Eukaryotic and bacterial DNA are not amplified by this kit.

Application

The PCR-based Venor GeM Mycoplasma Detection Kit employs PCR technology for rapid and reliable detection of mycoplasma DNA in cell cultures and virus stocks.[1][2]

Components

Does not include Taq Polymerase. Optimized for use with D9307, Taq DNA Polymerase

Storage and Stability

The kit components are stable during shipping at ambient temperature. Upon receipt, store at 2–8 °C. After rehydration of the primer/nucleotide mix, the positive control, and the internal control, store below –20 °C and avoid repeated freezing and thawing. For repeated testing of low sample numbers, primer/nucleotide mix and controls should be aliquoted after rehydration. By following these recommendations, the kit is stable until the expiration date stated on the label.

Legal Information

Venor is a trademark of Minerva Biolabs GmbH

Kit Components Only

Product No.
Description

  • Positive Control

  • Negative Control

  • PCR 10X Reaction Buffer 500 mL/vial

  • Primer/Nucleotide Mix 1 mL/vial

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anbarasu Kannan et al.
Scientific reports, 9(1), 11632-11632 (2019-08-14)
Triple negative breast cancer (TNBC) has poor survival, exhibits rapid metastases, lacks targeted therapies and reliable prognostic markers. Here, we examined metastasis promoting role of cancer testis antigen SPANXB1 in TNBC and its utility as a therapeutic target and prognostic
Enhanced neonatal Fc receptor function improves protection against primate SHIV infection.
Ko SY, Pegu A, Rudicell RS, et al.
Nature, 514(7524), 642-645 (2014)
Yanyan Li et al.
Oncotarget, 8(63), 106311-106323 (2018-01-02)
Decades of cytotoxic and more recently immunotherapy treatments for malignant glioma have had limited success due to dynamic intra-tumoral heterogeneity. The dynamic interplay of cancer cell subpopulations has been found to be under the control of proteins in the cancer
Mycoplasma detection in a historical arbovirus repository: Commercial kit comparison and implications for improved repository management
Russell BJ, et al.
Journal of Virological Methods (2020)
Tomohiro Aoki et al.
Cancer discovery, 10(3), 406-421 (2019-12-21)
Hodgkin lymphoma is characterized by an extensively dominant tumor microenvironment (TME) composed of different types of noncancerous immune cells with rare malignant cells. Characterization of the cellular components and their spatial relationship is crucial to understanding cross-talk and therapeutic targeting

Articles

Know when to use antibiotics to prevent bacterial or fungal, mycoplasma, or viral contamination in cell culture and find suitable antibiotics or other biological agents.

Know when to use antibiotics to prevent bacterial or fungal, mycoplasma, or viral contamination in cell culture and find suitable antibiotics or other biological agents.

Know when to use antibiotics to prevent bacterial or fungal, mycoplasma, or viral contamination in cell culture and find suitable antibiotics or other biological agents.

Know when to use antibiotics to prevent bacterial or fungal, mycoplasma, or viral contamination in cell culture and find suitable antibiotics or other biological agents.

See All

Protocols

Mycoplasma contamination of cell cultures is a serious issue impacting cell model validity. PCR testing for mycoplasma is an inexpensive, sensitive, and specific method for detecting contamination.

Questions

1–8 of 8 Questions  
  1. Can the cell culture supernatant be stored for a period of time before using the kit to determine if mycoplasma is present?

    1 answer
    1. The cell culture supernatant (sample) can be stored for several days at 4 °C before using the kit to determine if mycoplasma is present. For long-term storage, the samples can be stored at -20 °C, either in their native state or after heat inactivation.

      Helpful?

  2. Can I freeze the samples to collect several before using them?

    1 answer
    1. Samples should be cell supernatants when possible. Cell pellets are not recommended. The high concentration of DNA in the pellet is often responsible for the non-specific bands. If cell pellets are the only material available for testing, then centrifugation is necessary following sample boiling in order to remove all cell debris.

      Helpful?

  3. What is the difference between Product No. MP0025 (VenorGem Mycoplasma Detection Kit) and Product No. MP0035 (Lookout Mycoplasma Detection Kit)?

    1 answer
    1. The assay kits have a similar range of species detected (see product insert for listing).  For Product No. MP0025, the components are supplied as individual reagents. For Product No. MP0035, the components are supplied in reaction tubes, already mixed and ready for addition of sample and Taq. The internal control for Product No. MP0035 is at 481 bp rather than 191 bp as found in Product No. MP0025.

      Helpful?

  4. Can a cell lysate be used in the VenorGeM® Mycoplasma Kit, Product MP0025, assay?

    1 answer
    1. A cell lysate can contain PCR inhibitors.  Sigma Aldrich recommends that PCR inhibitors be removed using a DNA extraction with  commercially available kits. (Product Codes G1N10, G1N70, NA2000).

      Helpful?

  5. Product MP0025, VenorGeM® Mycoplasma Kit, does not contain TAQ.  What  DNA polymerase should be used?

    1 answer
    1. Any DNA polymerase with a 3’-5’ exonuclease activity is NOT recommended. These include Tli, Pfu,and Pwo. Those DNA polymerases that can be used include Taq, Tfl, and Tth. The kit has been optimized for use with Product No. D9307.

      Helpful?

  6. When using Product MP0025, VenorGeM® Mycoplasma Kit, there is inhibition in the sample (no internal control band).  Can the sample be diluted and the assay repeated?

    1 answer
    1. Sigma Aldrich does not recommend diluting the sample and re-assay as this can result in false negatives (sample diluted below the detection limits of the assay).  Sigma Aldrich recommends that PCR inhibitors be removed using a DNA extraction with  commercially available kits. (Product Codes G1N10, G1N70, NA2000).

      Helpful?

  7. The sample required for VenorGeM® Mycoplasma Kit, Product MP0025, is a cell culture supernatant.  Is this from adherent or suspension cells?

    1 answer
    1. The mycoplasma assay kit can be run on supernatants from either adherent or suspension cells. The centrifugation step during sample preparation will remove any cellular debris to limit PCR inhibitors.

      Helpful?

  8. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service