Skip to Content
Merck
All Photos(3)

Key Documents

C4790

Sigma-Aldrich

Carbenoxolone disodium salt

≥98%

Synonym(s):

3β-Hydroxy-11-oxoolean-12-en-30-oic acid 3-hemisuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C34H48O7Na2
CAS Number:
Molecular Weight:
614.72
EC Number:
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

powder

storage temp.

2-8°C

SMILES string

[Na].[H][C@@]12C[C@](C)(CC[C@]1(C)CC[C@]3(C)C2=CC(=O)[C@]4([H])[C@@]5(C)CC[C@H](OC(=O)CCC(O)=O)C(C)(C)[C@]5([H])CC[C@@]34C)C(O)=O

InChI

1S/C34H50O7.2Na/c1-29(2)23-10-13-34(7)27(32(23,5)12-11-24(29)41-26(38)9-8-25(36)37)22(35)18-20-21-19-31(4,28(39)40)15-14-30(21,3)16-17-33(20,34)6;;/h18,21,23-24,27H,8-17,19H2,1-7H3,(H,36,37)(H,39,40);;/q;2*+1/p-2/t21-,23?,24-,27+,30+,31?,32-,33+,34+;;/m0../s1

InChI key

BQENDLAVTKRQMS-FDPMUVMVSA-L

General description

Carbenoxolone disodium is a water soluble disodium salt of glycyrrhetinic acid hemisuccinate. It acts as an efficient promoter for the nasal absorption of insulin.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Promotion of nasal absorption of insulin by glycyrrhetinic acid derivatives. I.
Mishima M, et al.
Journal of Pharmacobio-Dynamics, 12(1), 31-36 (1989)
Seo-Yeon Yoon et al.
The journal of pain : official journal of the American Pain Society, 14(2), 205-214 (2013-02-05)
Spinal glial cells contribute to the development of many types of inflammatory and neuropathic pain. Here the contribution of spinal astrocytes and astrocyte gap junctions to oxaliplatin-induced mechanical hypersensitivity was explored. The expression of glial fibrillary acidic protein (GFAP) in
R A Warwick et al.
European journal of pain (London, England), 17(4), 571-580 (2012-10-16)
Chemotherapy-induced peripheral neuropathy is a serious side effect in cancer treatment, a major manifestation being neuropathic pain that can be debilitating and can reduce the quality of life of the patient. Oxaliplatin and taxol are common anti-cancer drugs that induce
Atsushi Sengiku et al.
Scientific reports, 8(1), 1996-1996 (2018-02-02)
Day-night changes in the storage capacity of the urinary bladder are indispensable for sound sleep. Connexin 43 (Cx43), a major gap junction protein, forms hemichannels as a pathway of ATP in other cell types, and the urinary bladder utilizes ATP
Jung-Sub Park et al.
Current biology : CB, 27(17), 2623-2629 (2017-08-22)
Topological distributions of individual cellular clocks have not been demonstrated in peripheral organs. The cochlea displays circadian patterns of core clock gene expression [1, 2]. PER2 protein is expressed in the hair cells and spiral ganglion neurons of the cochlea

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service