Merck
  • Multifunctional sequence-defined macromolecules for chemical data storage.
Multifunctional sequence-defined macromolecules for chemical data storage.
Nature communications (2018-10-28)
Steven Martens, Annelies Landuyt, Pieter Espeel, Bart Devreese, Peter Dawyndt, Filip Du Prez
ABSTRACT
Sequence-defined macromolecules consist of a defined chain length (single mass), end-groups, composition and topology and prove promising in application fields such as anti-counterfeiting, biological mimicking and data storage. Here we show the potential use of multifunctional sequence-defined macromolecules as a storage medium. As a proof-of-principle, we describe how short text fragments (human-readable data) and QR codes (machine-readable data) are encoded as a collection of oligomers and how the original data can be reconstructed. The amide-urethane containing oligomers are generated using an automated protecting-group free, two-step iterative protocol based on thiolactone chemistry. Tandem mass spectrometry techniques have been explored to provide detailed analysis of the oligomer sequences. We have developed the generic software tools Chemcoder for encoding/decoding binary data as a collection of multifunctional macromolecules and Chemreader for reconstructing oligomer sequences from mass spectra to automate the process of chemical writing and reading.
Sigma-Aldrich
2-Hydroxyethyl acrylate, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Isobornyl acrylate, technical grade, contains 200 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethylene glycol methyl ether acrylate, contains 50-100 ppm MEHQ as inhibitor, 98%
Sigma-Aldrich
Di(ethylene glycol) ethyl ether acrylate, technical grade, contains 1000 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Tetrahydrofurfuryl acrylate, contains 500 ppm hydroquinone as inhibitor, 500 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Propargyl acrylate, 98%
Sigma-Aldrich
2-(Dimethylamino)ethyl acrylate, contains <2,000 ppm MEHQ as inhibitor, 98%
Sigma-Aldrich
2-Chlorotrityl chloride, ≥97.0% (AT)
Sigma-Aldrich
Acetyl chloride solution, 1 M in methylene chloride