Skip to Content
Merck
All Photos(1)

Key Documents

26569

Sigma-Aldrich

2-Chlorotrityl chloride

≥97.0% (AT)

Synonym(s):

Chloro(2-chlorophenyl)diphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14Cl2
CAS Number:
Molecular Weight:
313.22
Beilstein:
1983914
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (AT)

form

solid

mp

130-135 °C

functional group

chloro
phenyl

SMILES string

Clc1ccccc1C(Cl)(c2ccccc2)c3ccccc3

InChI

1S/C19H14Cl2/c20-18-14-8-7-13-17(18)19(21,15-9-3-1-4-10-15)16-11-5-2-6-12-16/h1-14H

InChI key

JFLSOKIMYBSASW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chlorotrityl chloride was used to obtain protected peptide fragments that can be applied to the preparation of head-to-tail cyclopeptides or to condensation of peptidic fragments.

Other Notes

Protecting group reagent for carboxylic acids forming esters which are cleaved mildly.

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Barlos et al.
International journal of peptide and protein research, 37(6), 513-520 (1991-06-01)
The esterification of 2-chlorotrityl chloride resin with Fmoc-amino acids in the presence of DIEA is studied under various conditions. High esterification yields are obtained using 0.6 equiv. Fmoc-amino acid/mmol resin in DCM or DCE, in 25 min, at room temperature.
Shinichiro F Ichise et al.
Acta biomaterialia, 94, 361-371 (2019-06-15)
Collagen is the most abundant protein in the animal kingdom and has a unique triple-helical structure. It not only provides mechanical strength to tissues, but also performs specific biological functions as a multifaceted signaling molecule. Animal-derived collagen is therefore widely
Sogol Kangarlou et al.
Pharmaceutical biology, 55(1), 277-285 (2016-12-13)
Curcumin, a naturally occurring polyphenol, has been extensively studied for its broad-spectrum anticancer effects. The potential benefits are, however, limited due to its poor water solubility and rapid degradation which result in low bioavailability on administration. This study encapsulates curcumin
Jie Luo et al.
Journal of the American Society for Mass Spectrometry, 30(9), 1643-1653 (2019-06-07)
Chemical cross-linking combined with mass spectrometry (CL-MS) is a powerful method for characterizing the architecture of protein assemblies and for mapping protein-protein interactions. Despite its proven utility, confident identification of cross-linked peptides remains a formidable challenge, especially when the peptides
Asker Y Khapchaev et al.
Journal of peptide science : an official publication of the European Peptide Society, 22(11-12), 673-681 (2016-10-05)
Myosin light chain kinase (MLCK) is a key regulator of various forms of cell motility including smooth muscle contraction, cell migration, cytokinesis, receptor capping, secretion, etc. Inhibition of MLCK activity in endothelial and epithelial monolayers using cell-permeant peptide Arg-Lys-Lys-Tyr-Lys-Tyr-Arg-Arg-Lys (PIK

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service