Skip to Content
Merck
All Photos(1)

Key Documents

408913

Sigma-Aldrich

Ethylene glycol methyl ether acrylate

contains 50-100 ppm MEHQ as inhibitor, 98%

Synonym(s):

2-Methoxyethyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2CH2CH2OCH3
CAS Number:
Molecular Weight:
130.14
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

liquid

contains

50-100 ppm MEHQ as inhibitor

refractive index

n20/D 1.427 (lit.)

bp

56 °C/12 mmHg (lit.)

density

1.012 g/mL at 25 °C (lit.)

SMILES string

COCCOC(=O)C=C

InChI

1S/C6H10O3/c1-3-6(7)9-5-4-8-2/h3H,1,4-5H2,2H3

InChI key

HFCUBKYHMMPGBY-UHFFFAOYSA-N

General description

2-methoxyethyl acrylate can undergo RAFT copolymerization with itaconic anhydride.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Muta. 2 - Repr. 1B - Skin Corr. 1C - Skin Sens. 1 - STOT RE 2

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F

Flash Point(C)

60 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Daisuke Sato et al.
Physical chemistry chemical physics : PCCP, 22(38), 21672-21677 (2020-09-12)
Periodic patterns are ubiquitous in nature and spontaneously form on molecular to cosmic scales by the interplay between reaction and diffusion. Understanding how these patterns form is important to understand the construction rules of nature and apply them in the
RAFT copolymerization of itaconic anhydride and 2-methoxyethyl acrylate: a multifunctional scaffold for preparation of ?clickable? gold nanoparticles.
Javakhishvili I, et al.
Chemical Communications (Cambridge, England), 49, 4803-4805 (2013)
Luís Filipe-Ribeiro et al.
Food research international (Ottawa, Ont.), 129, 108855-108855 (2020-02-11)
4-Ethylphenol (4-EP) and 4-ethylguaiacol (4-EG) formation in red wines by Dekkera/Brettanomyces yeasts reduce significantly wine consumer's acceptability. Polymers with specific adsorption for volatile phenols (VPs) could be a valuable tool for wine producers for removing this negative sensory defect. In
Shunsuke Tazawa et al.
Macromolecular rapid communications, 41(19), e2000346-e2000346 (2020-08-19)
Thermoplastic solid poly(2-methoxyethyl acrylate) (PMEA)-based polyurethane (PU) is synthesized through the reversible addition-fragmentation chain transfer (RAFT) polymerization and the condensation polymerization, using hydroxyl-terminated RAFT reagents and diisocyanate, respectively. Neat PMEA is a promising antithrombogenic liquid used in the medical fields.
Thi Lien Nguyen et al.
Polymers, 12(8) (2020-08-19)
Biocompatible amphiphilic statistical copolymers P(MEA/MPC m

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service