Merck
Modeling the drug release from hydrogel-based matrices.
Molecular pharmaceutics (2014-12-17)
Diego Caccavo, Sara Cascone, Gaetano Lamberti, Anna Angela Barba
ABSTRACT
In this work the behavior of hydrogel-based matrices, the most widespread systems for oral controlled release of pharmaceuticals, has been mathematically described. In addition, the calculations of the model have been validated against a rich set of experimental data obtained working with tablets made of hydroxypropyl methylcellulose (a hydrogel) and theophylline (a model drug). The model takes into account water uptake, hydrogel swelling, drug release, and polymer erosion. The model was obtained as an improvement of a previous code, describing the diffusion in concentrated systems, and obtaining the erosion front (which is a moving boundary) from the polymer mass balance (in this way, the number of fitting parameters was also reduced by one). The proposed model was found able to describe all the observed phenomena, and then it can be considered a tool with predictive capabilities, useful in design and testing of new dosage systems based on hydrogels.
Product Description
Supelco
Theophylline, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Theophylline, anhydrous, ≥99%, powder
Theophylline, European Pharmacopoeia (EP) Reference Standard
USP
Theophylline, United States Pharmacopeia (USP) Reference Standard