Skip to Content
Merck
All Photos(1)

Key Documents

1392454

USP

Meropenem

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Meropenem trihydrate, (1R,5S,6S)-2-[(3S,5S)-5-(dimethylaminocarbonyl)pyrrolidin-3-ylthio]-6-[(R)-1-hydroxyethyl]-1-methylcarbapen-2-em-3-carboxylic acid trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H25N3O5S · 3H2O
CAS Number:
Molecular Weight:
437.51
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

meropenem

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

O.O.O.C[C@@H](O)[C@@H]1[C@H]2[C@@H](C)C(S[C@@H]3CN[C@@H](C3)C(=O)N(C)C)=C(N2C1=O)C(O)=O

InChI

1S/C17H25N3O5S.3H2O/c1-7-12-11(8(2)21)16(23)20(12)13(17(24)25)14(7)26-9-5-10(18-6-9)15(22)19(3)4;;;/h7-12,18,21H,5-6H2,1-4H3,(H,24,25);3*1H2/t7-,8-,9+,10+,11-,12-;;;/m1.../s1

InChI key

CTUAQTBUVLKNDJ-OBZXMJSBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Meropenem USP Reference standard intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monograph such as Meropenem for Injection

Biochem/physiol Actions

Meropenem trihydrate is an ultra-broad spectrum beta-lactam antibiotic active against both Gram-positive and Gram-negative bacteria.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ivan Barišić et al.
Infection, genetics and evolution : journal of molecular epidemiology and evolutionary genetics in infectious diseases, 27, 408-417 (2014-08-28)
The detailed analysis of antibiotic resistance mechanisms is essential for understanding the underlying evolutionary processes, the implementation of appropriate intervention strategies and to guarantee efficient treatment options. In the present study, 110 β-lactam-resistant, clinical isolates of Enterobacteriaceae sampled in 2011
A A Witney et al.
Clinical microbiology and infection : the official publication of the European Society of Clinical Microbiology and Infectious Diseases, 20(10), O609-O618 (2014-01-16)
A series of extensively drug-resistant isolates of Pseudomonas aeruginosa from two outbreaks in UK hospitals were characterized by whole genome sequencing (WGS). Although these isolates were resistant to antibiotics other than colistin, we confirmed that they are still sensitive to
Masaki Endo et al.
Plant & cell physiology, 56(1), 41-47 (2014-11-14)
The clustered regularly interspaced short palindromic repeat (CRISPR)-associated endonuclease 9 (CRISPR/Cas9) system has been demonstrated to be a robust genome engineering tool in a variety of organisms including plants. However, it has been shown that the CRISPR/Cas9 system cleaves genomic
Patrick N A Harris et al.
Trials, 16, 24-24 (2015-01-28)
Gram-negative bacteria such as Escherichia coli or Klebsiella spp. frequently cause bloodstream infections. There has been a worldwide increase in resistance in these species to antibiotics such as third generation cephalosporins, largely driven by the acquisition of extended-spectrum beta-lactamase or
Nicolò Girometti et al.
Medicine, 93(17), 298-309 (2014-11-15)
Multidrug resistance associated with extended-spectrum beta-lactamase (ESBL) and Klebsiella pneumoniae carbapenemase (KPC) among K. pneumoniae is endemic in southern Europe. We retrospectively analyzed the impact of resistance on the appropriateness of empirical therapy and treatment outcomes of K. pneumoniae bloodstream

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service