Skip to Content
Merck
All Photos(1)

Key Documents

1082708

USP

Butylated hydroxytoluene

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,6-Di-tert-butyl-4-methylphenol, 2,6-Di-tert-butyl-p-cresol, BHT, Butylated hydroxytoluene, Butylhydroxytoluene, DBPC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)3C]2C6H2(CH3)OH
CAS Number:
Molecular Weight:
220.35
Beilstein:
1911640
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor density

7.6 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

API family

butylated hydroxytoluene

autoignition temp.

878 °F

manufacturer/tradename

USP

bp

265 °C (lit.)

mp

69-73 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

Cc1cc(c(O)c(c1)C(C)(C)C)C(C)(C)C

InChI

1S/C15H24O/c1-10-8-11(14(2,3)4)13(16)12(9-10)15(5,6)7/h8-9,16H,1-7H3

InChI key

NLZUEZXRPGMBCV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Butylated hydroxytoluene USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Dutasteride and Tamsulosin Hydrochloride Capsules

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

260.6 °F - open cup

Flash Point(C)

127 °C - open cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Butylated Hydroxytoluene
USP43-NF38: United States Pharmacopeia and National Formulary
United States Pharmacopeia, 45(6), 5652-5652 (2021)
Toxicology and biochemistry of butylated hydroxyanisole and butylated hydroxytoluene.
A L Branen
Journal of the American Oil Chemists' Society, 52(2), 59-63 (1975-02-01)
Metabolism and pulmonary toxicity of butylated hydroxytoluene (BHT).
H Witschi et al.
Pharmacology & therapeutics, 42(1), 89-113 (1989-01-01)
A M Malkinson
Environmental mutagenesis, 5(3), 353-362 (1983-01-01)
Although the average American's daily consumption of BHT can be measured in milligrams, there are numerous reports that BHT causes organ damage in laboratory animals. Only a few genotoxic effects of BHT have been reported, however, including mutagenicity in the
Butylated hydroxytoluene (BHT): a review.
H Babich
Environmental research, 29(1), 1-29 (1982-10-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service