Skip to Content
Merck
All Photos(1)

Key Documents

07293

Supelco

tert-Butylhydroquinone

analytical standard

Synonym(s):

2-(1,1-Dimethylethyl)-1,4-benzenediol, NSC 4972

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CC6H3-1,4-(OH)2
CAS Number:
Molecular Weight:
166.22
Beilstein:
637923
EC Number:
MDL number:
UNSPSC Code:
12191503
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

autoignition temp.

855 °F

shelf life

limited shelf life, expiry date on the label

mp

127-129 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC(C)(C)c1cc(O)ccc1O

InChI

1S/C10H14O2/c1-10(2,3)8-6-7(11)4-5-9(8)12/h4-6,11-12H,1-3H3

InChI key

BGNXCDMCOKJUMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butylhydroquinone (TBHQ) is the primary metabolite of butylated hydroxyanisole. It is a widely used food antioxidant that acts by blocking free radicals to prevent the rancidity of oils and fats. TBHQ is also used as an antioxidant in cosmetic products.

Application

tert-Butylhydroquinone may be used as an analytical standard for the determination of the analyte in soybean biodiesel by differential pulse voltammetry (DPV) on glassy carbon (GC) electrode, and in oils, foods, biological fluids, and cosmetics by various chromatography techniques.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of antioxidants in cosmetics by micellar electrokinetic capillary chromatography with electrochemical detection
Guan Y, et al.
Journal of Chromatography A, 1074(1-2), 201-204 (2005)
Cytotoxicity and DNA damage properties of tert-butylhydroquinone (TBHQ) food additive
Eskandani M, et al.
Food Chemistry, 153(6), 315-320 (2014)
Quantitative determination of butylated hydroxyanisole, butylated hydroxytoluene, and tert-butyl hydroquinone in oils, foods, and biological fluids by high-performance liquid chromatography with fluorometric detection
Yankah VV, et al.
Lipids, 33(11), 1139-1145 (1998)
Development of an analytical method for the determination of tert-butylhydroquinone in soybean biodiesel
Goulart LA, et al.
Fuel: The Science and Technology of Fuel and Energy, 115(6), 126-131 (2014)
Tert-butylhydroquinone induces mitochondrial oxidative stress causing Nrf2 activation
Imhoff BR and Hansen JM
Cell Biology and Toxicology, 26(6), 541-551 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service