Skip to Content
Merck
All Photos(1)

Key Documents

1067704

USP

Betamethasone dipropionate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Betamethasone 17,21-dipropionate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[H][C@@]12C[C@H](C)[C@](OC(=O)CCl)(C(=O)COC(=O)CC)[C@@]1(C)C[C@H](O)C3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C27H34ClFO7/c1-5-22(33)35-14-21(32)27(36-23(34)13-28)15(2)10-19-18-7-6-16-11-17(30)8-9-24(16,3)26(18,29)20(31)12-25(19,27)4/h8-9,11,15,18-20,31H,5-7,10,12-14H2,1-4H3/t15-,18-,19-,20-,24-,25-,26?,27-/m0/s1

InChI key

UOCNBCVPZWLUKX-HOBFILNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Betamethasone 17,21-dipropionate is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sally A Helmy et al.
Therapeutic drug monitoring, 36(1), 100-107 (2013-11-26)
Diflunisal, naproxen, ketoprofen, etodolac, mefenamic acid, rofecoxib, and celecoxib are nonsteroidal anti-inflammatory drugs, which have analgesics, antipyretics, and anti-inflammatory activities. The aim of this work was to develop and validate a simple assay that could be implemented in most laboratories
Lawrence F Eichenfield et al.
Pediatric dermatology, 32(1), 28-35 (2014-11-22)
The objective of this study was to assess the safety and efficacy of the fixed combination calcipotriene 0.005% plus betamethasone dipropionate 0.064% topical suspension in adolescents with extensive scalp psoriasis. In this phase II, open-label, 8-week study, adolescents with psoriasis
Hanne Norsgaard et al.
Archives of dermatological research, 306(8), 719-729 (2014-07-17)
The calcipotriol/betamethasone dipropionate fixed-combination gel is widely used for topical treatment of psoriasis vulgaris. It has been hypothesized that calcipotriol counteracts glucocorticoid-induced skin atrophy which is associated with changes in the extracellular matrix (ECM). To elucidate the combined effects of
P C M van de Kerkhof et al.
Dermatology (Basel, Switzerland), 210(4), 294-299 (2005-06-09)
A two-compound product containing calcipotriol and betamethasone dipropionate (Daivobet/Dovobet) has been evaluated in a large clinical trial programme, providing a wealth of data on the treatment of psoriasis vulgaris. To determine the effectiveness of the two-compound product in patients with
Paul L McCormack
American journal of clinical dermatology, 12(6), 421-424 (2011-10-05)
Calcipotriene (calcipotriol)/betamethasone dipropionate (calcipotriene 50 μg/g and betamethasone 0.5 mg/g) is a fixed-dose combination of a vitamin D3 analog and a corticosteroid indicated for the once-daily, topical treatment of psoriasis vulgaris of the trunk, limbs, and scalp in adults. Both

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service