Skip to Content
Merck
All Photos(3)

Key Documents

C8037

Supelco

Clobetasol propionate

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32ClFO5
CAS Number:
Molecular Weight:
466.97
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98%

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CCl

InChI

1S/C25H32ClFO5/c1-5-21(31)32-25(20(30)13-26)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,27)19(29)12-23(18,25)4/h8-9,11,14,17-19,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1

InChI key

CBGUOGMQLZIXBE-XGQKBEPLSA-N

Gene Information

human ... NR3C1(2908)
rat ... Nr3c1(24413)

Looking for similar products? Visit Product Comparison Guide

General description

Clobetasol propionate is a novel and highly potent, topical corticosteroid, which is being assessed for its utility in the treatment of bullous pemphigoid.
Clobetasol propionate is a novel and highly potent, topical corticosteroid, which is being assessed for its utility in the treatment of bullous pemphigoid.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 4 - Repr. 1B - STOT RE 2

Target Organs

Adrenal gland,Immune system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Open preliminary clinical trial of clobetasol propionate ointment in adhesive paste for treatment of chronic oral vesiculoerosive diseases.
Lozada-Nur F and Huang M Z
Oral Surgery, Oral Medicine, Oral Pathology and Oral Radiology, 71(3), 283-287 (1991)
Open preliminary clinical trial of clobetasol propionate ointment in adhesive paste for treatment of chronic oral vesiculoerosive diseases
Lozada-Nur F, et al.
Oral Surgery, Oral Medicine, and Oral Pathology, 71, 283-287 (1991)
Treatment of bullous pemphigoid with topical clobetasol propionate
Westerhof W
Journal of the American Academy of Dermatology, 20, 458-461 (1989)
Treatment of bullous pemphigoid with topical clobetasol propionate.
Westerhof W.
Journal of the American Academy of Dermatology, 20(3), 458-461 (1989)
Mark G Lebwohl et al.
Journal of the American Academy of Dermatology, 69(3), 385-392 (2013-05-07)
Few clinical trials have evaluated the combination of topical corticosteroids plus systemic therapies for psoriasis. We sought to evaluate efficacy and safety of etanercept plus topical clobetasol propionate (CP) foam versus etanercept monotherapy for treatment of moderate to severe plaque

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service