Skip to Content
Merck
All Photos(1)

Key Documents

SAAR5885060

Sulfuric acid

uniLAB

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SO4
CAS Number:
Molecular Weight:
98.08
Beilstein:
2037554
MDL number:
UNSPSC Code:
12352322

grade

reagent

vapor density

<0.3 (25 °C, vs air)

vapor pressure

1 mmHg ( 146 °C)

product line

uniLAB

packaging

glass bottle of 1 × 2.5 L (SAAR5885060LC)
poly bottle of 1 × 2.5 L (SAAR5885060LP)
pkg of 25 L (SAAR5885060ME)

bp

~290 °C (lit.)

density

1.840 g/mL at 25 °C (lit.)

storage temp.

2-30°C

SMILES string

OS(O)(=O)=O

InChI

1S/H2O4S/c1-5(2,3)4/h(H2,1,2,3,4)

InChI key

QAOWNCQODCNURD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Chemically pure
For laboratory use only

Legal Information

uniLAB is a trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service