Skip to Content
Merck
All Photos(1)

Key Documents

P3817

Supelco

Phospholipid mixture for HPLC from Glycine max (soybean)

certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

biological source

soybean

Quality Level

grade

certified reference material

form

liquid

CofA

current certificate can be downloaded

packaging

pkg of 2 mL

concentration

in chloroform (varied)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

multi-component solution

functional group

phospholipid

shipped in

dry ice

storage temp.

−20°C

Application

Phospholipid mixture for HPLC from Glycine max (soybean) has been used as a standard in determining the total lipids extracted from the gills of blue mussels, Mytilus edulis via spotting them using thin layer chromatography (TLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    L-α-Lysophosphatidylcholine from Glycine max (soybean) .3 mg/mL
    L-α-Phosphatidylcholine 1.5 mg/mL
    L-α-Phosphatidylethanolamine from Glycine max (soybean) 1.2 mg/mL
    L-α-Phosphatidylinositol sodium salt from Glycine max (soybean) .9 mg/mL

related product

Product No.
Description
Pricing

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 2 - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3

Target Organs

Central nervous system, Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of seawater desalination and oil pollution on the lipid composition of blue mussels Mytilus edulis L. from the White Sea
Fokina N N, et al.
Ecotox. Environ. Safety, 110, 103-109 (2014)
Daniela Palgunow et al.
PloS one, 7(11), e46198-e46198 (2012-11-28)
Dietary restriction (DR) extends lifespan in man species and modulates evolutionary conserved signalling and metabolic pathways. Most of these studies were done in adult animals. Here we investigated fat phenotypes of C. elegans larvae and adults which were exposed to
Estela Area-Gomez et al.
The EMBO journal, 31(21), 4106-4123 (2012-08-16)
Alzheimer disease (AD) is associated with aberrant processing of the amyloid precursor protein (APP) by γ-secretase, via an unknown mechanism. We recently showed that presenilin-1 and -2, the catalytic components of γ-secretase, and γ-secretase activity itself, are highly enriched in
Marc D Tambini et al.
EMBO reports, 17(1), 27-36 (2015-11-14)
In addition to the appearance of senile plaques and neurofibrillary tangles, Alzheimer's disease (AD) is characterized by aberrant lipid metabolism and early mitochondrial dysfunction. We recently showed that there was increased functionality of mitochondria-associated endoplasmic reticulum (ER) membranes (MAM), a
Yi-Feng Kao et al.
Food & function, 9(7), 3986-3996 (2018-07-06)
The anti-inflammation properties of marine phospholipids enriched with n-3 fatty acids contribute to anti-inflammatory and inflammation-resolving mediators. Functional squid-skin (SQ) liposomes were manufactured from squid-skin phospholipids, and their anti-inflammatory effects were investigated. SQ liposomes included phosphatidylinositol (PI), phosphatidylserine (PS), phosphatidylethanolamine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service