Skip to Content
Merck
All Photos(1)

Key Documents

1787-1AMP

Supelco

Lipid Standard, Mono-, Di-, & Triglyceride Mix

certified reference material, pkg of 40 mg (Neat)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

pkg of 40 mg (Neat)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

functional group

ester

shipped in

dry ice

storage temp.

-10 to -25°C

Application

Lipid Standard, Mono-, Di-, & Triglyceride Mix was used in sample to analyse the lipid composition using TLC-FID system. It was also used as standard mix in analysing fractionated lipids using two-dimensional TLC.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    1,3-Diolein 10 mg
    1,2-Dioleoyl-rac-glycerol 10 mg
    Glyceryl trioleate 10 mg
    Monoolein 10 mg

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service