Skip to Content
Merck

CRM69091

Supelco

Dinotefuran

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

(RS)-N-Methyl-N′-nitro-N″-[(tetrahydro-3-furanyl)methyl]guanidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H14N4O3
CAS Number:
Molecular Weight:
202.21
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

Pricing and availability is not currently available.

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

SMILES string

CN\C(N[N+]([O-])=O)=N/CC1CCOC1

InChI

1S/C7H14N4O3/c1-8-7(10-11(12)13)9-4-6-2-3-14-5-6/h6H,2-5H2,1H3,(H2,8,9,10)

InChI key

YKBZOVFACRVRJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
62476L440873036
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

grade

for molecular biology

grade

for molecular biology

grade

-

grade

-

anion traces

sulfate (SO42-): ≤0.01%

anion traces

sulfate (SO42-): ≤50 mg/kg

anion traces

sulfate (SO42-): ≤0.01%

anion traces

sulfate (SO42-): ≤50 mg/kg

cation traces

Ba: ≤0.003%, K: ≤0.01%, heavy metals (as Pb): ≤0.002%

cation traces

Al: ≤5 mg/kg, As: ≤0.2 mg/kg, Ba: ≤10 mg/kg, Bi: ≤5 mg/kg, Ca: ≤10 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg

cation traces

Al: ≤0.0005%, Ba: ≤0.002%, Ca: ≤0.005%, Cu: ≤0.0005%, Fe: ≤0.002%, K: ≤0.005%, Mg: ≤0.005%, NH4+: ≤0.1%, Na: ≤0.005%, Pb: ≤0.001%, Zn: ≤0.001%

cation traces

Ba: ≤10 mg/kg, Ca: ≤20 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, K: ≤50 mg/kg, Mg: ≤5 mg/kg, Mn: ≤5 mg/kg, Na: ≤50 mg/kg, Ni: ≤10 mg/kg, Pb: ≤5 mg/kg, Zn: ≤5 mg/kg

concentration

8 M

concentration

-

concentration

-

concentration

-

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Dinotefuran is a third-generation chiral neonicotinoid insecticide, used against sucking, biting, and public health insects such as mosquitoes, ticks, beetles, aphids, whiteflies, and leaf hoppers. It disrupts the insect nervous system by exhibiting agonistic activity towards the nicotinic acetylcholine (ACh) receptors (nAChR) and affecting the cholinergic synaptic transmission.
Under the Regulation (EC) No 1107/2009 and revoking the Council Directives 79/117/EEC and 91/414/EEC, dinotefuran is not approved for its use as a plant protection agent in the European Union (EU). But a default maximum residue limit (MRL) of 0.01 mg/kg is set for dinotefuran in accordance with Art 18(1)(b) Reg 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
The dinotefuran CRM may also find given below uses:
  • Development of a method to simultaneously determine dinotefuran and its 3 polar metabolites in plum samples using dispersive-solid phase extraction procedure (d-SPE) followed by liquid chromatography-tandem mass spectrometry (LC-MS/MS)
  • Potentiometric determination of dinotefuran residues in soil and cucumber samples by using novel molecularly imprinted polymeric (MIP) material based sensors
  • Evaluation of an ultra high-performance liquid chromatography-tandem triple quadrupole mass spectrometry based method for the detection and quantification of dinotefuran and its metabolites in products of plant (watermelon, cucumber, rice) and animal (milk, egg, and pork) origin and also environmental samples of soil and water
  • Multi-residue analysis of pyridaben, dinotefuran, and metabolites of dinotefuran in soil and eggplant samples with a modified QuEChERS based sample extraction and rapid resolution liquid chromatography-triple quadrupole-tandem mass spectrometry (RRLC-QqQ-MS/MS)
  • Quantitative analysis of dinotefuran and its two metabolites in rice plant samples by high-performance liquid chromatography-tandem mass spectrometry (HPLC-MS/MS)
  • Development of a method to determine residues of dinotefuran and its two major metabolites in apple samples by QuEChERS based sample pre-treatment and liquid chromatography-tandem mass spectrometry (LC-MS/MS)

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Md Musfiqur Rahman et al.
Food chemistry, 239, 1235-1243 (2017-09-07)
A simple and effective method was developed for analyzing dinotefuran and its three metabolites (MNG, UF, and DN) in plum using liquid chromatography-tandem mass spectrometry (LC-MS/MS). Due to the polarity and high water miscibility, dinotefuran and some of its metabolites
Zenglong Chen et al.
Environment international, 130, 104854-104854 (2019-06-15)
In last decade, there has been a concerted effort to reduce the potential threats of honeybees' population due to exposure to neonicotinoid pesticides. A new perspective was put forward to reduce the potential ecological toxicity of neonicotinoid dinotefuran to honeybee
Xiaoxin Chen et al.
Chemosphere, 195, 245-251 (2017-12-23)
A sensitive method for simultaneous determination of pyridaben, dinotefuran, DN and UF in eggplant ecosystem was established and validated through rapid resolution liquid chromatography triples quadrupole tandem mass spectrometry (RRLC-QqQ-MS/MS). Matrix-matched external calibrations were introduced to check matrix effects. Limits
Weiwei Yu et al.
Environmental monitoring and assessment, 189(10), 521-521 (2017-09-28)
Residues of dinotefuran and its metabolites, 1-methyl-3-(tetrahydro-3-furylmethyl)urea (UF) and 1-methyl-3-(tetrahydro-3-furylmethyl)guanidine (DN), in apple were investigated using a "QuEChERS" (quick, easy, cheap, effective, rugged, safe) pretreatment and liquid chromatography-tandem mass spectrometry. Limits of detection (LODs) and quantification (LOQs) of dinotefuran, UF
Zenglong Chen et al.
Environment international, 130, 104854-104854 (2019-06-15)
In last decade, there has been a concerted effort to reduce the potential threats of honeybees' population due to exposure to neonicotinoid pesticides. A new perspective was put forward to reduce the potential ecological toxicity of neonicotinoid dinotefuran to honeybee

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service