CRM30806
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[g]-2-benzopyran
certified reference material, TraceCERT®, mixture of isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylindeno[5,6-c]pyran, Hexamethylindanopyran
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
shelf life
limited shelf life, expiry date on the label
manufacturer/tradename
Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
storage temp.
−20°C
SMILES string
CC1(C)C(C)C(C)(C)C2=CC3=C(C(C)COC3)C=C21
InChI
1S/C18H26O/c1-11-14-10-16-15(9-13(14)7-8-19-11)17(3,4)12(2)18(16,5)6/h9-12H,7-8H2,1-6H3
InChI key
YWMTVUUYOAIDBC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma aldrich.com
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Flash Point(F)
291.2 °F - Pensky-Martens closed cup
Flash Point(C)
144 °C - Pensky-Martens closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service