Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[
g
]-2-benzopyran
Synonym(s):
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylindeno[5,6-
c
]pyran, Hexamethylindanopyran
Empirical Formula (Hill Notation):
C
18
H
26
O
CAS Number:
1222-05-5
Molecular Weight:
258.40
EC Number:
214-946-9
Products
Brand
Supelco (2)
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
certified reference material (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
CRM30806
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[
g
]-2-benzopyran,
certified reference material,
Trace
CERT
®
, mixture of isomers, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Expand
View Pricing
57524
1,3,4,6,7,8-Hexahydro-4,6,6,7,8,8-hexamethylcyclopenta[
g
]-2-benzopyran,
analytical standard
Expand
View Pricing