Skip to Content
Merck
All Photos(1)

Key Documents

CRM06772

Supelco

Chlorobromouron

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

Chlorbromuron

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H10BrClN2O2
CAS Number:
Molecular Weight:
293.54
Beilstein:
2128736
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

storage temp.

2-8°C

SMILES string

CON(C)C(=O)Nc1ccc(Br)c(Cl)c1

InChI

1S/C9H10BrClN2O2/c1-13(15-2)9(14)12-6-3-4-7(10)8(11)5-6/h3-5H,1-2H3,(H,12,14)

InChI key

NLYNUTMZTCLNOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com
Chlorbromouron is an obsolete systemic phenylurea herbicide used for pre- and post- emergence control of annual grasses (e.g. foxtail) and broad leafed weeds (e.g. chickweed) on a variety of crops. It inhibits photosynthesis by interfering with the electron transfer in photosystem II. As per the commission regulation no 1107/2009, repealing the council directive 91/414/EEC, chlorobromouron is not approved for use in the European Union. A default maximum residue limit (MRL) of 0.01 mg/kg is set for chlorbromuron as per regulation (EC) no. 396/2005.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques.
Chlorobromouron CRM can also be used as below:

  • Method development for the quantification of 25 polar pesticides in fruit, vegetable, and water samples using high-performance liquid chromatography(HPLC)
  • Separation and determination of 12 substituted phenylurea herbicides by HPLC in the study to assess the leaching of the herbicides in two agricultural soil samples
  • Development of an HPLC method with diode array detection following dispersive liquid-liquid microextraction to determine 11 phenylurea herbicides in natural waters
  • Investigation of 1,6-hexanediol ethoxylate diacrylate (HEDA)-based polymeric monoliths as a sorbent for the extraction of phenylurea herbicides from water samples
  • Multi-residue analysis of 250 pesticides in surface water samples using liquid chromatography- quadrupole-Orbitrap high resolution tandem mass spectrometry

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive liquid-liquid microextraction for the determination of herbicides of urea derivatives family in natural waters by HPLC
Amelin VG, et al.
Journal of Analytical Chemistry, 68(9), 822- 830 (2013)
Shu-Ling Lin et al.
Talanta, 147, 199-206 (2015-11-26)
In this study, recently developed 1,6-hexanediol ethoxylate diacrylate (HEDA)-based polymeric monoliths were utilized as sorbents for efficient extraction of phenylurea herbicides (PUHs) from water samples. The HEDA-based monolithic sorbents were prepared in a fused silica capillary (0.7mm i.d., 4.5-cm long)
Determination of polar pesticides in water, vegetables, and fruits by high performance liquid chromatography
Amelin V G, et al.
Moscow University Chemistry Bulletin, 67(6), 275- 282 (2012)
Abiotic and biotic processes governing the fate of phenylurea herbicides in soils: a review
Hussain S, et al.
Critical reviews in environmental science and technology, 45(18), 1947- 1998 (2015)
Simón Navarro et al.
Journal of agricultural and food chemistry, 60(21), 5279-5286 (2012-05-15)
In this study, the potential groundwater pollution of 12 substituted phenylurea herbicides (chlorbromuron, chlorotoluron, diuron, fenuron, fluometuron, isoproturon, linuron, metobromuron, metoxuron, monolinuron, Monuron, and neburon) was investigated under laboratory conditions. For this purpose, leaching studies were conducted using disturbed soil

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service