Skip to Content
Merck
All Photos(2)

Key Documents

57308

Supelco

SPME fiber assembly Polydimethylsiloxane (PDMS)

greener alternative

df 30 μm(PDMS, needle size 24 ga, for use with manual holder

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115854

material

fused silica fiber
plain yellow hub

Quality Level

needle size

24 ga

packaging

pkg of 3 ×  ea

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

df

100 μm (PDMS)
30 μm (PDMS)
7 μm (PDMS)

technique(s)

solid phase microextraction (SPME): suitable

fiber L

1 cm

matrix active group

PDMS coating

application(s)

food and beverages

compatibility

for analyte group volatiles (MW 80-500)
for use with manual holder

greener alternative category

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.

Application

Nonpolar Semivolatiles

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

609.8 °F - closed cup

Flash Point(C)

321 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid-Phase Microextraction for Cannabinoids Analysis in Hair and it Possible Application to Other Drugs
Strano-Rossi, S., et al.
Journal of Analytical Toxicology, 23 (1999)
B J Hall et al.
Analytical chemistry, 70(9), 1788-1796 (1998-05-26)
Solid-phase microextraction (SPME) is applied to the determination of cannabidiol, delta 8-tetrahydrocannabinol (delta 8-THC), delta 9-tetrahydrocannabinol (delta 9-THC), and cannabinol in pure water and human saliva. The inherent extraction behavior of the cannabinoids in pure water is evaluated along with
R M Tomaino et al.
Journal of agricultural and food chemistry, 49(8), 3993-3998 (2001-08-22)
To evaluate the impact of Cheddar cheese starter cultures on the level of free fatty acids in liquid whey, a solid-phase microextraction (SPME) technique was utilized. The determination of response factors relative to an internal standard and the verification of
Alejandra Delgado et al.
Journal of separation science, 31(4), 768-774 (2008-02-02)
Headspace solid-phase microextraction was optimised for the simultaneous preconcentration of methylmercury (MeHg+), monobutyltin, dibutyltin, tributyltin, monophenyltin (MPhT), diphenyltin (DPhT), and triphenyltin (TPhT) from sediments and biota. Extraction time (3-24 min), extraction temperature (20-90 degrees C), desorption time (1-10.4 min), desorption
S Niedziella et al.
Journal of chromatography. A, 885(1-2), 457-464 (2000-08-15)
Solid-phase microextraction using a 30 microns polydimethylsiloxane fibre has been used to sample the volatile organic compounds from standard mixtures and from mixtures produced by the decomposition of organic compounds. This method of sampling has been compared with the direct

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service