508489
Crimp seals with PTFE/silicone septa
aluminum seal (open center), diam. × thickness 20 mm × 2.9 mm, PTFE/silicone (blue silicone), Chromacol 20-AC-ST3, pkg of 100 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
PTFE/silicone (blue silicone)
aluminum seal (open center)
packaging
pkg of 100 ea
manufacturer/tradename
Chromacol 20-AC-ST3
O.D.
20 mm
diam. × thickness
20 mm × 2.9 mm
fitting
thread for 20 mm
Related Categories
General description
It is used as an air-tight seal, and prevents contamintion
related product
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service