Skip to Content
Merck
All Photos(1)

Key Documents

50166-U

Nucleosil® C18 HPLC Column

5 μm particle size, L × I.D. 25 cm × 4 mm

Synonym(s):

C18 Nucleosil column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115700
eCl@ss:
32110501
NACRES:
SB.52

Agency

suitable for USP L1

Quality Level

product line

Nucleosil®

feature

endcapped

manufacturer/tradename

Nucleosil®

extent of labeling

15% Carbon loading

parameter

≤70 °C temp. range
400 bar pressure (5801 psi)

technique(s)

HPLC: suitable

L × I.D.

25 cm × 4 mm

surface area

350 m2/g

matrix

octadecyl

matrix active group

C18 (octadecyl) phase

particle size

5 μm

pore size

100 Å

separation technique

reversed phase

Looking for similar products? Visit Product Comparison Guide

Application



  • HPLC analysis of mitoxantrone in mouse plasma and tissues: application in a pharmacokinetic study.: This study demonstrates the application of HPLC using a Nucleosil® C18 column for analyzing mitoxantrone levels in mouse plasma and tissues, providing valuable data for pharmacokinetic evaluations (An et al., 2010).


  • Determination of pKa values of nonsteroidal anti-inflammatory drug-oxicams by RP-HPLC and their analysis in pharmaceutical dosage forms.: This research utilizes the Nucleosil® C18 HPLC column to determine the pKa values of oxicams, aiding in the quality control of pharmaceutical formulations (Demiralay et al., 2009).


  • Development of an analytical method for the determination of beta2-agonist residues in animal tissues by high-performance liquid chromatography with on-line electrogenerated [Cu(HIO6)2]5- -luminol chemiluminescence detection.: The study describes a method using a Nucleosil® C18 column to detect beta2-agonist residues, showcasing its sensitivity and application in food safety analysis (Zhang et al., 2007).


  • Assay of artemether, methylparaben, and propylparaben in a formulated pediatric antimalarial dry suspension.: Utilizing the Nucleosil® C18 column, this study establishes an assay for key components in pediatric antimalarial formulations, contributing to better quality control (Atemnkeng et al., 2007).


Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Nucleosil is a registered trademark of Macherey-Nagel GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service