Skip to Content
Merck
All Photos(1)

Key Documents

49451-U

Supelco

C7 - C30 Saturated Alkanes

certified reference material, 1000 μg/mL each component in hexane

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

1000 μg/mL each component in hexane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

petroleum

format

multi-component solution

storage temp.

2-8°C

General description

Quantitative standard suitable for use as a retention index marker probe.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Analyte

Description

    Decane
    Docosane
    Dodecane
    Eicosane
    Heneicosane
    Heptacosane
    Heptadecane
    Hexacosane
    Hexadecane
    Heptane
    Nonacosane
    Nonadecane
    Nonane
    Octacosane
    Octadecane
    Octane
    Pentacosane
    Pentadecane
    Tetracosane
    Tetradecane
    Triacontane
    Tricosane
    Tridecane
    Undecane
    See All (24)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F

Flash Point(C)

-22 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fang Yuan et al.
Metabolites, 10(9) (2020-08-29)
Untargeted metabolomics is expected to lead to a better mechanistic understanding of diseases and thus applications of precision medicine and personalized intervention. To further increase metabolite coverage and achieve high accuracy of metabolite quantification, the present proof-of-principle study was to
C M S Ambrosio et al.
Journal of applied microbiology, 129(3), 541-553 (2020-04-10)
This study investigated the in vitro mechanism of action of a commercial citrus EO, Brazilian orange terpenes (BOT), on an enterotoxigenic Escherichia coli (ETEC) isolated from pig gut and on Lactobacillus rhamnosus. Firstly, bacteria were exposed sequentially to BOT every
Chemical characterization of three accessions of Brassica juncea L. extracts from different plant tissues
Oulad El Majdoub Y, et al.
Molecules (Basel), 25(22), 5421-5421 (2020)
Gabriel Goetten de Lima et al.
Ecotoxicology and environmental safety, 180, 616-623 (2019-05-28)
Pyroligneous acid (PA) is a by-product of bio-oil, which is obtained by pyrolysis of the wood. This product has been tested for use in several areas, such as agriculture, as a promising green herbicide; however, there are few scientific data
Robert J Arnold et al.
PloS one, 14(8), e0220787-e0220787 (2019-08-09)
The whiskey industry is dominated by whiskey styles with recipes that contain corn as the primary grain. However, little research has been conducted to investigate whiskey specific distinctions arising from different corn varieties and growing environments (i.e. terroir). Further, no

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service