Skip to Content
Merck
All Photos(1)

Key Documents

48490

Supelco

Benzo[b]fluoranthene

analytical standard

Synonym(s):

2,3-Benzfluoranthene, 3,4-Benz[e]acephenanthrylene, 3,4-Benzfluoranthene, 3,4-Benzofluoranthene, Benz[e]acephenanthrylene, Benzo[e]fluoranthene, NSC 89265

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H12
CAS Number:
Molecular Weight:
252.31
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 50 mg

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

163-165 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

c1ccc-2c(c1)-c3cccc4c3c-2cc5ccccc45

InChI

1S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12H

InChI key

FTOVXSOBNPWTSH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benzo[b]fluoranthene is a polycyclic aromatic hydrocarbon, which belongs to the benzofluoranthene family obtained via incomplete combustion of gasoline engine exhaust, coal and oil combustion products, broiled and smoked foods.

Application

Benzo[b]fluoranthene may be used as an analytical standard for the determination of the analyte in meat and fish products, smoked and non-smoked black teas and tea infusions and water samples by chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Determination of polycyclic aromatic hydrocarbons in smoked and non-smoked black teas and tea infusions
Pincemaille J, et al.
Food Chemistry, 145, 807-813 (2014)
Determination of polycyclic aromatic hydrocarbons in water samples by solid phase extraction and gas chromatography
Kanchanamayoon W and Tatrahun N
World Journal of chemistry, 3(2), 51-54 (2008)
Benzo [b] fluoranthene: tumorigenicity in strain A/J mouse lungs, DNA adducts and mutations in the Ki-ras oncogene
Maa.JM, et al.
Carcinogenesis, 17, 1701-1704 (1996)
The determination of polycyclic aromatic hydrocarbons in smoked fish by gas chromatography mass spectrometry with positive-ion chemical ionization
Yurchenko S and Molder U
Journal of food composition and analysis, 18(8), 857-869 (2005)
Determination of fluoranthene, benzo [b] fluoranthene and benzo [a] pyrene in meat and fish products and their intake by Malaysian
Jahurul M, et al.
Food Additives and Contaminants, 1, 73-80 (2013)

Articles

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

This application note describes the fast and efficient separation of the EU’s list of 15 + 1 polynuclear aromatic hydrocarbons (PAHs) using Ascentis® Express PAH HPLC column.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service